Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IVTQWQPRN CDPMVDFLDS WVHIIPVWIL DNILDQLIFP KLQKEVENWN PLTDTVPIHS WIHPWLPLMQ ARLEPLYSPI RSKLSSALQK WHPSDSSAKL ILQPWKDVFT PGSWEAFMVK NIVPKLGMCL GELVINPHQQ HMDAFYWVID WEGMISVSSL VGLLEKHFFP KWLQVLCSWL SNSPNYEEIT KWYLGWKSMF SDQVLAHPSV KDKFNEALDI MNRAVSSNVG AYMQPGAREN IAYLTHTERR KDFQYEAMQE RREAENMAQR GIGVAASSVP MNFKDLIETK AEEHNIVFMP VIGKRHEGKQ LYTFGRIVIY IDRGVVFVQG EKTWVPTSLQ SLIDMAK (492-837 aa encoded by BC033080 )
Involved in pre-mRNA splicing, specifically in spliceosome disassembly during late-stage splicing events. Intron turnover seems to proceed through reactions in two lariat-intron associated complexes termed Intron Large (IL) and Intron Small (IS). In cooperation with DHX15 seems to mediate the transition of the U2, U5 and U6 snRNP-containing IL complex to the snRNP-free IS complex leading to efficient debranching and turnover of excised introns. May play a role in the differentiation of ameloblasts and odontoblasts or in the forming of the enamel extracellular matrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Septin and tuftelin-interacting protein 1; STIP-1; Tuftelin-interacting protein 11
Gene Aliases: bK445C9.6; hNtr1; HSPC006; NTR1; Spp382; STIP; TFIP11; TIP39
UniProt ID: (Human) Q9UBB9
Entrez Gene ID: (Human) 24144
Molecular Function:
RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support