Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
TGF-beta1, -2, and -3 are a closely related group of proteins (70-80% sequence homology) that are produced by many cell types and function as growth and differentiation factors. The active forms of TGF-beta1, -2, and -3 are disulfide-linked homodimers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BSC-1 cell growth inhibitor; Cetermin; G-TSF; Glioblastoma-derived T-cell suppressor factor; latency-associated peptide; MGF; Milk growth factor; polyergin; prepro-transforming growth factor beta-1; prepro-transforming growth factor beta-2; prepro-transforming growth factor beta-3; regulatory protein; tgf beta 1; TGF beta 2 protein; tgf beta 3; tgf beta1; TGF-beta 1; TGF-beta-1; TGF-beta-2; TGF-beta-3; TGF-beta3; Transforming growth factor; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; Transforming growth factor beta-2; Transforming growth factor beta-2 proprotein; Transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor beta-3-like protein; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta 2; transforming growth factor, beta 3; transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta-1 precursor
Gene Aliases: ARVD; ARVD1; BB105277; CED; DPD1; I79_000092; LAP; LDS4; LDS5; MGF; RNHF; TGF-B2; TGF-B3; TGF-beta1; TGF-beta2; TGF-beta3; TGFB; Tgfb-1; Tgfb-2; Tgfb-3; TGFB1; TGFB2; TGFB3; TGFbeta; TGFbeta1; TGFbeta2; TGFbeta3
UniProt ID: (Human) P01137, (Bovine) P18341, (Human) P61812, (Bovine) P21214, (Human) P10600, (Rat) P17246, (Mouse) P04202, (Mouse) P27090, (Rat) Q07257, (Rat) Q07258
Entrez Gene ID: (Human) 7040, (Dog) 403998, (Bovine) 282089, (Human) 7042, (Bovine) 534069, (Dog) 488596, (Human) 7043, (Bovine) 538957, (Dog) 490796, (Rat) 59086, (Chinese hamster) 100773359, (Mouse) 21803, (Chinese hamster) 100774918, (Mouse) 21808, (Rat) 81809, (Rat) 25717, (Mouse) 21809, (Chinese hamster) 100756831
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support