Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • TGFBR2 Antibodies

            Invitrogen

            TGFBR2 Polyclonal Antibody

            1 Published Figure
            1 Reference
            View all (31) TGFBR2 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Catalog # PA5-88257
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            487.00
            100 µL
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite TGFBR2 Polyclonal Antibody

            • Antibody Testing Data (2)
            • Published Figures (1)
            Application
            TGFBR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 3

            TGFBR2 Antibody (PA5-88257) in IHC (P)

            Immunohistochemistry analysis of TGFBR2 in paraffin-embedded mouse spleen. Samples were incubated with TGFBR2 Polyclonal antibody (Product # PA5-88257) using a dilution of 1:100 (40x lens). Perform high pressure antigen retrieval with 10 mM citrate buffer pH 6.0 before commencing with IHC staining protocol.
            View Product

            TGFBR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            TGFBR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            TGFBR2 Antibody in Immunohistochemistry (IHC)

            Product Details

            PA5-88257

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:1,000
            -

            Immunohistochemistry (IHC)

            -
            View 1 publication 1 publication

            Immunohistochemistry (Paraffin) (IHC (P))

            1:50-1:200
            -

            Immunocytochemistry (ICC/IF)

            1:50-1:200
            -

            ELISA (ELISA)

            1 µg/mL
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Human

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Recombinant fusion protein containing a sequence corresponding to amino acids 23-280 of human TGF beta Receptor II (TGFBR2) (NP_0032334)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.82 mg/mL

            Purification

            Affinity Chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.05% ProClin 300

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2804781

            Product Specific Information

            Positive Samples: Mouse spleen, Rat lung; Cellular Location: Cell membrane, Single-pass type I membrane protein

            Immunogen sequence: TIPPHVQKSV NNDMIVTDNN GAVKFPQLCK FCDVRFSTCD NQKSCMSNCS ITSICEKPQE VCVAVWRKND ENITLETVCH DPKLPYHDFI LEDAASPKCI MKEKKKPGET FFMCSCSSDE CNDNIIFSEE YNTSNPDLLL VIFQVTGISL LPPLGVAISV IIIFYCYRVN RQQKLSSTWE TGKTRKLMEF SEHCAIILED DRSDISSTCA NNINHNTELL PIELDTLVGK GRFAEVYKAK LKQNTSEQFE TVAVKIFP

            Target Information

            Transforming Growth Factor, beta receptor 2 (AAT3, FAA3, MFS2, RIIC, LDS1B, LDS2B, TAAD2, TGFR-2, TGFbeta-RII) is a member of the TGF-beta family of receptor Serine/Threonine kinases. It is involved in the regulation of cell proliferation. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). TGFBR2 encodes the gamma subunit of the catalytic core. Alternatively spliced transcript variants encoding different isoforms have been identified. TGFBR2 also has a pseudogene on chromosome 14.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: tbetaR-II; tgf beta receptor 2; TGF-beta receptor II; TGF-beta receptor type II; TGF-beta receptor type IIB; TGF-beta receptor type-2; TGF-beta type II receptor; TGFbeta RII; TGFR-2; TGFR2; transforming growth factor beta receptor II; transforming growth factor beta receptor type II; transforming growth factor beta receptor type IIC; transforming growth factor, beta receptor II; transforming growth factor, beta receptor II (70/80kDa); transforming growth factor, beta receptor II alpha; transforming growth factor, beta receptor II beta; transforming growth factor, beta receptor II delta; transforming growth factor, beta receptor II epsilon; transforming growth factor, beta receptor II gamma; transforming growth factor, beta receptor IIT; transforming growth factor-b type II receptor; Transforming growth factor-beta receptor type II; transforming growth factor-beta type II receptor

            View more View less

            Gene Aliases: 1110020H15Rik; AAT3; AU042018; DNIIR; FAA3; LDS1B; LDS2; LDS2B; MFS2; RIIC; RIIDN; TAAD2; TbetaR-II; TbetaRII; TBR-II; TGF-beta 2; TGFbeta-RII; TGFBR2; Tgfbr2T; TGFR-2

            View more View less

            UniProt ID: (Human) P37173, (Rat) P38438, (Mouse) Q62312

            View more View less

            Entrez Gene ID: (Human) 7048, (Rat) 81810, (Mouse) 21813

            View more View less

            Function(s)

            transmembrane receptor protein serine/threonine kinase activity receptor signaling protein serine/threonine kinase activity transforming growth factor beta-activated receptor activity transforming growth factor beta receptor activity, type II protein binding ATP binding glycosaminoglycan binding mitogen-activated protein kinase kinase kinase binding type I transforming growth factor beta receptor binding SMAD binding metal ion binding transforming growth factor beta binding type III transforming growth factor beta receptor binding protein homodimerization activity protein heterodimerization activity nucleotide binding protein kinase activity protein serine/threonine kinase activity receptor activity kinase activity transferase activity transferase activity, transferring phosphorus-containing groups serine/threonine protein kinase receptor

            Process(es)

            blood vessel development patterning of blood vessels vasculogenesis in utero embryonic development heart looping positive regulation of mesenchymal cell proliferation lens development in camera-type eye positive regulation of tolerance induction to self antigen positive regulation of B cell tolerance induction positive regulation of T cell tolerance induction outflow tract morphogenesis growth plate cartilage chondrocyte growth protein phosphorylation receptor-mediated endocytosis apoptotic process transforming growth factor beta receptor signaling pathway common-partner SMAD protein phosphorylation Notch signaling pathway smoothened signaling pathway gastrulation brain development heart development embryo implantation aging response to nutrient positive regulation of cell proliferation response to mechanical stimulus response to glucose regulation of gene expression positive regulation of epithelial cell migration positive regulation of epithelial to mesenchymal transition peptidyl-serine phosphorylation peptidyl-threonine phosphorylation signal transduction by protein phosphorylation negative regulation of transforming growth factor beta receptor signaling pathway organ regeneration activation of protein kinase activity embryonic hemopoiesis wound healing regulation of cell proliferation response to drug myeloid dendritic cell differentiation positive regulation of skeletal muscle tissue regeneration response to estrogen positive regulation of angiogenesis response to steroid hormone digestive tract development positive regulation of smooth muscle cell proliferation embryonic cranial skeleton morphogenesis positive regulation of NK T cell differentiation palate development negative regulation of cardiac muscle cell proliferation pathway-restricted SMAD protein phosphorylation ventricular septum morphogenesis bronchus morphogenesis trachea formation mammary gland morphogenesis lung lobe morphogenesis response to cholesterol positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation lens fiber cell apoptotic process positive regulation of reactive oxygen species metabolic process response to hypoxia growth plate cartilage development transmembrane receptor protein serine/threonine kinase signaling pathway negative regulation of cell proliferation organ morphogenesis response to organic substance response to organic cyclic compound lung development cartilage development lung morphogenesis bronchus development trachea morphogenesis phosphorylation cell differentiation regulation of growth
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0