Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Peptide sequence: RILPDMLQQR RNDPIIGHKT GKDAHATHLQ STEASVPAKS GPVVQTMYKA
Sequence homology: Human: 100%; Yeast: 90%
TGIF2LY is a member of the TALE/TGIF homeobox family of transcription factors. This gene lies within the male specific region of chromosome Y, in a block of sequence that is thought to be the result of a large X-to-Y transposition.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein TGIF2LY; TGF-beta-induced transcription factor 2-like protein; TGFB-induced factor 2-like protein, Y-linked; TGFB-induced factor 2-like, Y-linked; TGFB-induced factor homeobox 2-like, Y-linked; TGIF-like on the Y
Gene Aliases: TGIF2LY; TGIFLY
UniProt ID: (Human) Q8IUE0
Entrez Gene ID: (Human) 90655
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support