Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDQVATLRLE SVDLQSSRNN KEHHTQEMGV KRLTVRRGQP FYLRLSFSRP FQSQNDHITF VAETGPKPSE LLGTRATFFL TRVQPGNVWS ASDFTIDSNS LQVSLFTPAN AVIGHYTLKI EISQGQGHSV TYPLGTFILL FNPWSPEDDV YLPSEILLQE YIMRDYGFVY KGHERFITSW PWNYGQFEED IIDICFEILN KSLYHLKNPA KDCSQRNDVV YVCRVVSAMI NSNDDNGVLQ GNWGEDYSKG VSPLEWKGSV AILQQWSARG GQPVKYGQCW VFASVMCTVM RCLGVPTRVV SNFRSAHNVD RNLTIDTYYD RNAEMLSTQK RDKIWNFHVW NECWMIRKDL PPGYNGWQVL DPTPQQTSSG LFCCGPASVK AIREGDVHLA YDTPFVYAEV NADEVIWLLG DGQAQEILAH NTSSIGKEIS TKMVGSDQRQ SITSSYKYPE GSPEERAVFM KASRKMLGPQ RASLPFLDLL ESGGLRDQPA QLQLHLARIP EWGQDLQLLL RIQRVPDSTH PRGPIGLVVR FCAQALLHGG GTQKPFWRHT VRMNLDFGKE TQWPLLLPYS NYRNKLTDEK LIRVSGIAEV EETGRSMLVL KDICLEPPHL SIEVSERAEV GKALRVHVTL TNTLMVALSS CTMVLEGSGL INGQIAKDLG TLVAGHTLQI QLDLYPTKAG PRQLQVLISS NEVKEIKGYK DIFVTVAGAP
Transglutaminases (TGM; EC 2.3.2.13) are a family of structurally and functionally related enzymes that stabilize protein assemblies through the formation of gamma-glutamyl-epsilon lysine crosslinks. For additional background information on transglutaminases, see TGM1 (MIM 190195).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein-glutamine gamma-glutamyltransferase Z; TG(Z); TGase Z; TGase-7; TGMZEC 2.3.2.13; TGZ; Transglutaminase Z; Transglutaminase-7
Gene Aliases: TGM7; TGMZ
UniProt ID: (Human) Q96PF1
Entrez Gene ID: (Human) 116179
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support