Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: SVKQEEAGVR PSAGNVSTHP SLSQRPGGST KSHPEPQTPK DSPSKSSAEA QTPEDTPNKS GAEAKTQKDS SNKSGAEAKT QKGSTSKSGS EAQTTKDSTS KSHPELQTPK DSTGKSGAEA QTPEDSPNRS GAEAKTQKDS PSKSGSEAQT TKDVPNKSGA DGQTPKDGSS KSGAEDQTPK DVPNKSGAEK QTPKDGSNKS GAEEQGPIDG PSKSGAEEQT SKDSPNKVVP EQPSRKDHSK PISNPSDNKE LPKADTNQLA DKGKLSPHAF KTESGEETDL ISPPQEEVKS SEPTEDVEPK EAEDDDTGPE EGSPPKEEKE KMSGSASSEN REGTLSDSTG SEKDDLYPNG SGNGSAESSH
IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53; MIM 191170), whereas ASPP1. and ASPP2. are activators of p53.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hTGN48; hTGN51; TGN38 homolog; Trans-Golgi network glycoprotein 46; Trans-Golgi network glycoprotein 48; Trans-Golgi network glycoprotein 51; Trans-Golgi network integral membrane protein 2; trans-Golgi network protein (46, 48, 51kD isoforms); Trans-Golgi network protein 2; trans-Golgi network protein TGN51
Gene Aliases: TGN38; TGN46; TGN48; TGN51; TGOLN2; TTGN2
UniProt ID: (Human) O43493
Entrez Gene ID: (Human) 10618
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support