Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRVIAIDID PVKIALARNN AEVYGIADKI EFICGDFLLL ASFLKADVVF LSPPWGGPDY ATAETFDIRT MMSPDGFEIF RLSKKITNNI VYFLPRNADI DQVASLAGPG GQVEIEQNFL NNKLKTITAY FGDLIRRPAS ET (1-141 aa encoded by BC011999)
Catalyzes the 2 serial methylation steps for the conversion of the 7-monomethylguanosine (m(7)G) caps of snRNAs and snoRNAs to a 2,2,7-trimethylguanosine (m(2,2,7)G) cap structure. The enzyme is specific for guanine, and N7 methylation must precede N2 methylation. Hypermethylation of the m7G cap of U snRNAs leads to their concentration in nuclear foci, their colocalization with coilin and the formation of canonical Cajal bodies (CBs). Plays a role in transcriptional regulation. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cap-specific guanine-N2 methyltransferase; CLL-associated antigen KW-2; Hepatocellular carcinoma-associated antigen 137; nuclear receptor coactivator 6 interacting protein; Nuclear receptor coactivator 6-interacting protein; PIMT; PIPMT; PRIP-interacting protein with methyltransferase motif; Trimethylguanosine synthase
Gene Aliases: D4Ertd800e; HCA137; NCOA6IP; PIMT; PIPMT; TGS1
UniProt ID: (Human) Q96RS0, (Mouse) Q923W1
Entrez Gene ID: (Human) 96764, (Mouse) 116940
Molecular Function:
RNA methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support