Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTRSCSAVGC STRDTVLSRE RGLSFHQFPT DTIQRSKWIR AVNRVDPRSK KIWIPGPGAI LCSKHFQESD FESYGIRRKL KKGAVPSVSL YKIPQGVHLK GKARQKILKQ PLPDNSQEVA TEDHNYSLKT PLTIGAEKLA EVQQMLQVSK KRLISVKNYR MIKKRKGLRL IDALVEEKLL SEETECLLRA QFSDFKWELY NWRETDEYSA EMKQFACTLY LCSSKVYDYV RKILKLPHSS ILRTWLSKCQ PSPGFNSNIF SFLQRRVENG DQLYQYCSLL IKSMPLKQQL QWDPSSHSLQ GFMDFGLGKL DADETPLASE TVLLMAVGIF GHWRTPLGYF FVNRASGYLQ AQLLRLTIGK LSDIGITVLA VTSDATAHSV QMAKALGIHI DGDDMKCTFQ HPSSSSQQIA YFFDSCHLLR LIRNAFQNFQ SIQFINGIAH WQHLVELVAL EEQELSNMER IPSTLANLKN HVLKVNSATQ LFSESVASAL EYLLSLDLPP FQNCIGTIHF LRLINNLFDI FNSRNCYGKG LKGPLLPETY SKINHVLIEA KTIFVTLSDT SNNQIIKGKQ KLGFLGFLLN AESLKWLYQN YVFPKVMPFP YLLTYKFSHD HLELFLKMLR QVLVTSSSPT CMAFQKAYYN LETRYKFQDE VFLSKVSIFD ISIARRKDLA LWTVQRQYGV SVTKTVFHEE GICQDWSHCS LSEALLDLSD HRRNLICYAG YVANKLSALL TCEDCITALY ASDLKASKIG SLLFVKKKNG LHFPSESLCR VINICERVVR THSRMAIFEL VSKQRELYLQ QKILCELSGH INLFVDVNKH LFDGEVCAIN HFVKLLKDII ICFLNIRAKN VAQNPLKHHS ERTDMKTLSR KHWSSVQDYK CSSFANTSSK FRHLLSNDGY PFK
THAP9 (THAP domain-containing protein 9) is a 903 amino acid protein that contains one THAP-type zinc finger. The gene that encodes THAP9 contains roughly 19,448 bases and maps to human chromosome 4q21.22. Chromosome 4 represents approximately 6% of the human genome and contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease, is on chromosome 4. FGFR-3 is also encoded on chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer. Chromosome 4 is also tied to Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA transposase THAP9; hTh9; THAP domain-containing protein 9
Gene Aliases: hTh9; THAP9
UniProt ID: (Human) Q9H5L6
Entrez Gene ID: (Human) 79725
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support