Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PSYLDKDEQ CVVCGDKATG YHYRCITCEG CKGFFRRTIQ KNLHPTYSCK YDSCCVIDKI TRNQCQLCRF KKCIAVGMAM DLVLDDSKRV AKRKLIEQNR ERRRKEEMIR SLQQRPEPTP EEWDLIHIAT EAHRSTNAQG SHWKQRRKFL PDDIGQSPIV SMPDGDKVDL EAFSEFTKII TPAITRVVDF AKKLPMFSEL PCEDQIILLK GCCMEIMSLR AAVRYDPESD TLTLSGEMAV (44-282 aa encoded by BC002728)
The protein encoded by this gene is a nuclear hormone receptor for triiodothyronine. It is one of the several receptors for thyroid hormone, and has been shown to mediate the biological activities of thyroid hormone. Knockout studies in mice suggest that the different receptors, while having certain extent of redundancy, may mediate different functions of thyroid hormone. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1; c-erb-A thyroid hormone receptor; c-erbA-1; C-erbA-alpha; c-erbA-alpha2; c-erbAalpha; c-erbAalpha2; cerbA protein; cerbAalpha; E ERB-T-1; EAR-7; ERBA ALPHA; ERBA related 7; ERBA-related 7; MGC000261; MGC43240; Nuclear receptor subfamily 1 group A member 1; Oncogene ERBA; Thyroid hormone receptor alpha; thyroid hormone receptor alpha 1; thyroid hormone receptor alpha 1 (avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1 formerly ERBA1); thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a) oncogene homolog, avian); thyroid normone nuclear receptor alpha variant 1; TR alpha 1; TR alpha 1+2; TR alpha 2; triiodothyronine receptor; V-erbA-related protein 7
Gene Aliases: 6430529J03Rik; AR7; AW259572; c-ERBA-1; C-erba-alpha; CHNG6; EAR7; ERB-T-1; ERBA; ERBA1; NR1A1; Rvr; T3R[a]; T3Ralpha; THRA; THRA1; THRA2
UniProt ID: (Human) P10827, (Mouse) P63058, (Rat) P63059
Entrez Gene ID: (Human) 7067, (Mouse) 21833, (Rat) 81812
Molecular Function:
C4 zinc finger nuclear receptor
DNA-binding transcription factor
zinc finger transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support