Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene contains a type 1 thrombospondin domain, which is found in a number of proteins involved in the complement pathway, as well as in extracellular matrix proteins. Alternatively spliced transcript variants encoding different isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4833423O18Rik; Thrombospondin type-1 domain-containing protein 1; thrombospondin, type I, domain 1; thrombospondin, type I, domain containing 1; Transmembrane molecule with thrombospondin module
Gene Aliases: 4833423O18Rik; THSD1; TMTSP; UNQ3010; UNQ3010/PRO9769
UniProt ID: (Human) Q9NS62, (Mouse) Q9JM61
Entrez Gene ID: (Human) 55901, (Rat) 364630, (Mouse) 56229
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support