Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Probable protein tyrosine-kinase transmembrane receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: tyrosine kinase receptor 1; tyrosine kinase with immunoglobulin and epidermal growth factor homology domains 1; tyrosine kinase with immunoglobulin-like and EGF-like domains 1; Tyrosine-protein kinase receptor Tie-1
Gene Aliases: D430008P04Rik; JTK14; TIE; Tie-1; TIE1
UniProt ID: (Human) P35590, (Mouse) Q06806
Entrez Gene ID: (Human) 7075, (Mouse) 21846, (Rat) 89806
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support