Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
FIGURE: 1 / 1
Immunogen sequence: QMGLADSVKD FQRKGTHIVL WTGDQELELQ RLFEEFRDSD DVLGHIMKNI TAKRSRARIV DKLLALGLVA ERRELYKKRQ KKLASSILPN GAESLKDFCQ EDLEEEENLP EEDSEEEEEG GSEAEQVQGS LVLSNENLGQ SLHQEGFSIL LLWLQNCLIR AADDREEDGC SQAVPLVPLT EENEEAMENE QFQQLLRKLG VRPPASGQET FWRIPAKLSP TQLRRAAASL SQPEEEQKLQ PELQPKVPGE QGSDEEHCKE HRAQALRALL LAHKKKAGLA SPEEEDAVGK EPLKAAPKKR QLLDSDEEQE EDEGRNRAPE LGAPGIQKKK RYQIEDDEDD
The general transcription factor IIE (TFIIE) is part of the RNA polymerase II transcription initiation complex, recruiting TFIIH and being essential for promoter clearance by RNA polymerase II. TFIIE is a heterodimer (and sometimes heterotetramer) of alpha and beta subunits. The protein encoded by this gene represents the beta subunit of TFIIE.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hTIM; mTim; Protein timeless homolog; rTIM; rTLP; timeless circadian clock 1; timeless homolog; timeless {ECO:0000312|RGD:620939}; Timeless-like protein; Tof1 homolog
Gene Aliases: C77407; Debt69; hTIM; TIM; TIM1; TIMELESS; TIMELESS1
UniProt ID: (Human) Q9UNS1, (Mouse) Q9R1X4, (Rat) Q9Z2Y1
Entrez Gene ID: (Human) 8914, (Mouse) 21853, (Rat) 83508
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support