Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDPLRAQQL AAELEVEMMA DMYNRMTSAC HRKCVPPHYK EAELSKGESV CLDRCVSKYL DIHERMGKKL TELSMQDEEL MKRVQQSSGP A (1-90 aa encoded by BC032133)
The mitochondrial protein encoded by this gene belongs to a family of evolutionarily conserved proteins that are organized in heterooligomeric complexes in the mitochondrial intermembrane space. These proteins mediate the import and insertion of hydrophobic membrane proteins into the mitochondrial inner membrane, functioning as intermembrane space chaperones for the highly insoluble carrier proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial import inner membrane translocase subunit Tim10; small zinc finger-like protein; translocase of inner mitochondrial membrane 10 homolog; translocase of inner mitochondrial membrane 13 homolog a
Gene Aliases: TIM10; TIM10A; Tim13; TIMM10; Timm13a
UniProt ID: (Human) P62072, (Rat) P62074, (Mouse) P62073
Entrez Gene ID: (Human) 26519, (Rat) 64464, (Mouse) 30059
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support