Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEEYAREPC PWRIVDDCGG AFTMGVIGGG VFQAIKGFRN APVGIRHRLR GSANAVRIRA PQIGGSFAVW GGLFSTIDCG LVRLRGKEDP WNSITSGALT GAVLAARSGP LAMVGSAMMG GILLALIEGV GILLTRYTAQ QFRNAPPFLE DPSQLPPKDG TPAPGYPSYQ QYH (1-172 aa encoded by BC010142)
This gene encodes a multipass transmembrane protein that forms an integral component of the mitochondrial translocase TIM23 complex. This complex facilitates the transport of mitochondrial proteins from the cytosol across the mitochondrial inner membrane and into the mitochondrion. There is a pseudogene for this gene on chromosome 12. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: inner mitochondrial membrane preprotein translocase; Mitochondrial import inner membrane translocase subunit Tim17-B; translocator of inner mitochondrial membrane 17 kDa, b; translocator of inner mitochondrial membrane 17b; translocator of inner mitochondrial membrane b
Gene Aliases: 17kDa; DXS9822; JM3; mTim17b; RGD1561744; Sfc3; TIM17B; TIMM17B
UniProt ID: (Human) O60830, (Mouse) Q9Z0V7
Entrez Gene ID: (Human) 10245, (Mouse) 21855, (Rat) 317374
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support