Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Positive Samples: 293T, HepG2, Mouse liver, Mouse brain
Immunogen sequence: MAASAAVFSR LRSGLRLGSR GLCTRLATPP RRAPDQAAEI GSRGSTKAQG PQQQPGSEGP SYAKKVALWL AGLLGAGGTV SVVYIFGNNP VDENGAKIPD
TIMM50 (Mitochondrial import inner membrane translocase subunit TIM50) is an enzyme that in humans is encoded by the TIMM50 gene. Tim50 is a subunit of the Tim23 translocase complex in the inner mitochondrial membrane. Mutations in TIMM50 can lead to epilepsy, severe intellectual disability, and 3-methylglutaconic aciduria. TIMM50 expression is increased in breast cancer cells and decreased in hypertrophic hearts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homolog of yeast Tim50; MGC102733; Mitochondrial import inner membrane translocase subunit TIM50; similar to translocase of inner mitochondrial membrane 50 homolog; Tim50-like protein; translocase of inner mitochondrial membrane 50 homolog
Gene Aliases: 2810403L02Rik; AU015082; PRO1512; TIM50; TIM50L; TIMM50
UniProt ID: (Human) Q3ZCQ8, (Mouse) Q9D880
Entrez Gene ID: (Rat) 687295, (Human) 92609, (Mouse) 66525
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support