Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TM7SF2 is involved in the conversion of lanosterol to cholesterol.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-beta-hydroxysterol Delta (14)-reductase; Another new gene 1 protein; C-14 sterol reductase; C14SR; delta(14)-sterol reductase; Delta(14)-sterol reductase TM7SF2; Delta-14-SR; Putative sterol reductase SR-1; Sterol C14-reductase; Transmembrane 7 superfamily member 2
Gene Aliases: 3110041O18Rik; ANG1; C14SR; Dhcr14; DHCR14A; NET47; TM7SF2
UniProt ID: (Human) O76062, (Mouse) Q71KT5
Entrez Gene ID: (Human) 7108, (Mouse) 73166, (Rat) 293688
Molecular Function: reductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support