Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TM9SF3 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dinucleotide oxidase disulfide thiol exchanger 3 superfamily member 3; endomembrane protein emp70 precursor isolog; EP70-P-iso; P70-P-iso; RP11-34E5.1; SM-11044 binding protein; SM-11044-binding protein; Transmembrane 9 superfamily member 3; Transmembrane 9 superfamily member 3-like protein; transmembrane protein 9 superfamily member 3; transmembrane protein TM9SF3
Gene Aliases: 1810073M23Rik; 2810031D16Rik; AI115521; AI413748; AW146116; AW549777; EP70-P-iso; fb75h12; mKIAA4036; RGD1564625; SMBP; TM9SF3; UNQ245/PRO282; wu:fa94f05; wu:fb75h12; zgc:56246
UniProt ID: (Human) Q9NWL8, (Mouse) Q9ET30
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support