Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of total TMBIM1.
Negatively regulates aortic matrix metalloproteinase-9 (MMP9) production and may play a protective role in vascular remodeling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein lifeguard 3; Protein RECS1; Protein RECS1 homolog; Responsive to centrifugal force and shear stress gene 1 protein; Transmembrane BAX inhibitor motif-containing protein 1
Gene Aliases: 2310061B02Rik; AA960455; AU024746; C78899; LFG3; mKIAA4161; MST100; MSTP100; PP1201; PSEC0158; RECS1; Tmbib1; TMBIM1
UniProt ID: (Human) Q969X1, (Mouse) Q8BJZ3
Entrez Gene ID: (Human) 64114, (Mouse) 69660, (Rat) 316516
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support