Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene is considered a member of a gene family predicted to encode transmembrane proteins. The specific function of this gene is unknown; however, expression in the inner ear suggests that it may be crucial for normal auditory function. Mutations in this gene may underlie hereditary disorders of balance and hearing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4933416G09Rik; Beethoven; Bth; CWEA1; dJ686C3.3; dn; transmembrane channel-like 2; transmembrane channel-like gene family 2; Transmembrane channel-like protein 2; transmembrane cochlear-expressed gene 2; Transmembrane cochlear-expressed protein 2; transmembrane, cochlear expressed 2
Gene Aliases: C20orf145; CWEA2; TMC2; UNQ907/PRO1928
UniProt ID: (Human) Q8TDI7, (Mouse) Q8R4P4
Entrez Gene ID: (Human) 117532, (Rat) 296153, (Mouse) 192140
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support