Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GETEKKCFIE EIPDETMVIG NYRTQLYDKQ REEYQPATPG LGMFVEVKDP EDKVILARQY GSEGRFTFTS HTPGEHQICL HS
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
TMED9 (also named as GMP25, GP25L2 or p25) is a member of the EMP24/GP25L family. TMED9 probably forms hetero-oligomeric complexes with TMED7 (p27), TMED2 (p24), and TMED10 (p23). Located in the endoplasmic reticulum and the Golgi, TMED9 appears to be involved in vesicular protein trafficking, mainly in the early secretory pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glycoprotein 25L2; GMP25; gp25L2 protein; p24 family protein alpha-2; p24alpha2; p25; Transmembrane emp24 domain-containing protein 9; transmembrane emp24 protein transport domain containing 9
Gene Aliases: GMP25; GP25L2; HSGP25L2G; p24a2; p25; RGD1307627; TMED9
UniProt ID: (Human) Q9BVK6, (Rat) Q5I0E7
Entrez Gene ID: (Human) 54732, (Rat) 361207
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support