Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 77%; Horse: 93%; Human: 100%; Mouse: 79%
TMEM132B is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Transmembrane protein 132B
Gene Aliases: KIAA1786; KIAA1906; TMEM132B
UniProt ID: (Human) Q14DG7
Entrez Gene ID: (Human) 114795
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support