Product References
NETs Promote Inflammatory Injury by Activating cGAS-STING Pathway in Acute Lung Injury.
International journal of molecular sciences
Zhao J,Zhen N,Zhou Q,Lou J,Cui W,Zhang G,Tian B
Tue Mar 07 00:00:00 UTC 2023
The BET PROTAC inhibitor dBET6 protects against retinal degeneration and inhibits the cGAS-STING in response to light damage.
Journal of neuroinflammation
Zhu X,Liu W,Tang X,Chen Y,Ge X,Ke Q,Liang X,Gan Y,Zheng Y,Zou M,Deng M,Liu Y,Li DW,Gong L
Mon May 22 00:00:00 UTC 2023
Glutamylation of an HIV-1 protein inhibits the immune response by hijacking STING.
Cell reports
Qian G,Zhang Y,Liu Y,Li M,Xin B,Jiang W,Han W,Wang Y,Tang X,Li L,Zhu L,Sun T,Yan B,Zheng Y,Xu J,Ge B,Zhang Z,Yan D
Tue May 30 00:00:00 UTC 2023
Aβ Induces Neuroinflammation and Microglial M1 Polarization via cGAS-STING-IFITM3 Signaling Pathway in BV-2 Cells.
Neurochemical research
Wu Z,Tang W,Ibrahim FEEM,Chen X,Yan H,Tao C,Wang Z,Guo Y,Fu Y,Wang Q,Ge Y
Fri Sep 01 00:00:00 UTC 2023
STING/TBK1 Regulates Inflammation in Macrophages and Titanium Particles-Induced Osteolysis.
ACS biomaterials science & engineering
Ouyang Z,Xu J,Liu T,Lin S,Sun Y,Huang Y,Zheng Z,Zeng G,Li C,Li S,Ding Y
Mon Jun 12 00:00:00 UTC 2023
Soluble CD4 effectively prevents excessive TLR activation of resident macrophages in the onset of sepsis.
Signal transduction and targeted therapy
Zhang SY,Xu QP,Shi LN,Li SW,Wang WH,Wang QQ,Lu LX,Xiao H,Wang JH,Li FY,Liang YM,Gong ST,Peng HR,Zhang Z,Tang H
Mon Jun 19 00:00:00 UTC 2023
Soluble CD4 effectively prevents excessive TLR activation of resident macrophages in the onset of sepsis.
Signal transduction and targeted therapy
Zhang SY,Xu QP,Shi LN,Li SW,Wang WH,Wang QQ,Lu LX,Xiao H,Wang JH,Li FY,Liang YM,Gong ST,Peng HR,Zhang Z,Tang H
Mon Jun 19 00:00:00 UTC 2023
Rhinovirus C replication is associated with the endoplasmic reticulum and triggers cytopathic effects in an in vitro model of human airway epithelium.
PLoS pathogens
Gagliardi TB,Goldstein ME,Song D,Gray KM,Jung JW,Ignacio MA,Stroka KM,Duncan GA,Scull MA
Sat Jan 01 00:00:00 UTC 2022
Lipotoxicity-induced STING1 activation stimulates MTORC1 and restricts hepatic lipophagy.
Autophagy
Liu K,Qiu D,Liang X,Huang Y,Wang Y,Jia X,Li K,Zhao J,Du C,Qiu X,Cui J,Xiao Z,Qin Y,Zhang Q
Fri Apr 01 00:00:00 UTC 2022
Hydrogen Sulfide Ameliorated High Choline-Induced Cardiac Dysfunction by Inhibiting cGAS-STING-NLRP3 Inflammasome Pathway.
Oxidative medicine and cellular longevity
Bai L,Dai J,Xia Y,He K,Xue H,Guo Q,Tian D,Xiao L,Zhang X,Teng X,Wu Y,Jin S
Tue Aug 02 00:00:00 UTC 2022
LPS-Induced Activation of the cGAS-STING Pathway is Regulated by Mitochondrial Dysfunction and Mitochondrial DNA Leakage in Endometritis.
Journal of inflammation research
Li MZ,Wen XY,Liu XQ,Wang YQ,Yan L
Sat Oct 15 00:00:00 UTC 2022
LPS-Induced Activation of the cGAS-STING Pathway is Regulated by Mitochondrial Dysfunction and Mitochondrial DNA Leakage in Endometritis.
Journal of inflammation research
Li MZ,Wen XY,Liu XQ,Wang YQ,Yan L
Sat Oct 15 00:00:00 UTC 2022
LPS-Induced Activation of the cGAS-STING Pathway is Regulated by Mitochondrial Dysfunction and Mitochondrial DNA Leakage in Endometritis.
Journal of inflammation research
Li MZ,Wen XY,Liu XQ,Wang YQ,Yan L
Sat Oct 15 00:00:00 UTC 2022
LPS-Induced Activation of the cGAS-STING Pathway is Regulated by Mitochondrial Dysfunction and Mitochondrial DNA Leakage in Endometritis.
Journal of inflammation research
Li MZ,Wen XY,Liu XQ,Wang YQ,Yan L
Sat Oct 15 00:00:00 UTC 2022
LPS-Induced Activation of the cGAS-STING Pathway is Regulated by Mitochondrial Dysfunction and Mitochondrial DNA Leakage in Endometritis.
Journal of inflammation research
Li MZ,Wen XY,Liu XQ,Wang YQ,Yan L
Sat Oct 15 00:00:00 UTC 2022
LPS-Induced Activation of the cGAS-STING Pathway is Regulated by Mitochondrial Dysfunction and Mitochondrial DNA Leakage in Endometritis.
Journal of inflammation research
Li MZ,Wen XY,Liu XQ,Wang YQ,Yan L
Sat Oct 15 00:00:00 UTC 2022
Curcumol Suppresses CCF-Mediated Hepatocyte Senescence Through Blocking LC3B-Lamin B1 Interaction in Alcoholic Fatty Liver Disease.
Frontiers in pharmacology
Qi X,Zheng S,Ma M,Lian N,Wang H,Chen L,Song A,Lu C,Zheng S,Jin H
Sat Jul 16 00:00:00 UTC 2022
AMICA1 is a diagnostic and prognostic biomarker and induces immune cells infiltration by activating cGAS-STING signaling in lung adenocarcinoma.
Cancer cell international
Feng Z,Zhang Y,He M,Han Y,Cai C,Liu S,Liu P,Chen Y,Shen H,Zeng S
Sat Mar 05 00:00:00 UTC 2022
AMICA1 is a diagnostic and prognostic biomarker and induces immune cells infiltration by activating cGAS-STING signaling in lung adenocarcinoma.
Cancer cell international
Feng Z,Zhang Y,He M,Han Y,Cai C,Liu S,Liu P,Chen Y,Shen H,Zeng S
Sat Mar 05 00:00:00 UTC 2022
Human STING Is Regulated by an Autoinhibitory Mechanism for Type I Interferon Production.
Journal of innate immunity
Qi S,Wang C,Zhang R,Zhu J,Hou X,Jiang Y,Li J,Ren M,Li M,Hou F
Thu Sep 08 00:00:00 UTC 2022
Therapeutic targeting of STING-TBK1-IRF3 signalling ameliorates chronic stress induced depression-like behaviours by modulating neuroinflammation and microglia phagocytosis.
Neurobiology of disease
Duan N,Zhang Y,Tan S,Sun J,Ye M,Gao H,Pu K,Wu M,Wang Q,Zhai Q
Fri Jul 01 00:00:00 UTC 2022
Macrophage SCAP Contributes to Metaflammation and Lean NAFLD by Activating STING-NF-κB Signaling Pathway.
Cellular and molecular gastroenterology and hepatology
Huang X,Yao Y,Hou X,Wei L,Rao Y,Su Y,Zheng G,Ruan XZ,Li D,Chen Y
Mon Jun 20 00:00:00 UTC 2022
Macrophage SCAP Contributes to Metaflammation and Lean NAFLD by Activating STING-NF-κB Signaling Pathway.
Cellular and molecular gastroenterology and hepatology
Huang X,Yao Y,Hou X,Wei L,Rao Y,Su Y,Zheng G,Ruan XZ,Li D,Chen Y
Mon Jun 20 00:00:00 UTC 2022
Macrophage SCAP Contributes to Metaflammation and Lean NAFLD by Activating STING-NF-κB Signaling Pathway.
Cellular and molecular gastroenterology and hepatology
Huang X,Yao Y,Hou X,Wei L,Rao Y,Su Y,Zheng G,Ruan XZ,Li D,Chen Y
Mon Jun 20 00:00:00 UTC 2022
Macrophage SCAP Contributes to Metaflammation and Lean NAFLD by Activating STING-NF-κB Signaling Pathway.
Cellular and molecular gastroenterology and hepatology
Huang X,Yao Y,Hou X,Wei L,Rao Y,Su Y,Zheng G,Ruan XZ,Li D,Chen Y
Mon Jun 20 00:00:00 UTC 2022
Inhibition of cGAS-STING by JQ1 alleviates oxidative stress-induced retina inflammation and degeneration.
Cell death and differentiation
Zou M,Ke Q,Nie Q,Qi R,Zhu X,Liu W,Hu X,Sun Q,Fu JL,Tang X,Liu Y,Li DW,Gong L
Thu Sep 01 00:00:00 UTC 2022
GBP3 promotes glioblastoma resistance to temozolomide by enhancing DNA damage repair.
Oncogene
Xu H,Jin J,Chen Y,Wu G,Zhu H,Wang Q,Wang J,Li S,Grigore FN,Ma J,Chen CC,Lan Q,Li M
Fri Jul 01 00:00:00 UTC 2022
TBK1 recruitment to STING activates both IRF3 and NF-κB that mediate immune defense against tumors and viral infections.
Proceedings of the National Academy of Sciences of the United States of America
Yum S,Li M,Fang Y,Chen ZJ
Tue Apr 06 00:00:00 UTC 2021
The DJ1-Nrf2-STING axis mediates the neuroprotective effects of Withaferin A in Parkinson's disease.
Cell death and differentiation
Zhao M,Wang B,Zhang C,Su Z,Guo B,Zhao Y,Zheng R
Sun Aug 01 00:00:00 UTC 2021
Staphylococcus aureus Induces IFN-β Production via a CARMA3-Independent Mechanism.
Pathogens (Basel, Switzerland)
Zhou Y,Zhao S,Gao X,Jiang S,Ma J,Wang R,Li Q,Qin L,Tong Z,Wu J,Zhao J
Thu Mar 04 00:00:00 UTC 2021
Golgi apparatus-synthesized sulfated glycosaminoglycans mediate polymerization and activation of the cGAMP sensor STING.
Immunity
Fang R,Jiang Q,Guan Y,Gao P,Zhang R,Zhao Z,Jiang Z
Tue May 11 00:00:00 UTC 2021
Golgi apparatus-synthesized sulfated glycosaminoglycans mediate polymerization and activation of the cGAMP sensor STING.
Immunity
Fang R,Jiang Q,Guan Y,Gao P,Zhang R,Zhao Z,Jiang Z
Tue May 11 00:00:00 UTC 2021
STING agonist and IDO inhibitor combination therapy inhibits tumor progression in murine models of colorectal cancer.
Cellular immunology
Shi J,Liu C,Luo S,Cao T,Lin B,Zhou M,Zhang X,Wang S,Zheng T,Li X
Sun Aug 01 00:00:00 UTC 2021