Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is rat brain tissue lysate.
PA5-20931 can be used with blocking peptide PEP-1045.
TMEM184A, a multi-pass transmembrane protein, belongs to the UPF0206 family. While the exact function of TMEM184A remains unclear, it is known to be expressed at high levels in the developing testis. TMEM184A may mediate sex-specific signaling events during Sertoli cell differentiation. The restricted expression of this gene in Sertoli cells suggests a role for the encoded protein in the development and/or continued function of that cell type. Ectopic expression of TMEM184A in female genital ridges resulted in failure of gonocytes to enter meiosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sexually dimorphic expressed in male gonads 1; tcag7.540; Transmembrane protein 184A
Gene Aliases: RGD1306702; Sdmg1; TMEM184A
UniProt ID: (Human) Q6ZMB5, (Mouse) Q3UFJ6, (Rat) Q4QQS1
Entrez Gene ID: (Human) 202915, (Mouse) 231832, (Rat) 304325
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support