Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NTQHKFTSG RVAFQLLQVG VDSPAAGTRH NSAGPAHLHR ALSTASSMQR LPAATRATLI LSLAFASLHS ACSAASSSNS SSLTAHHPDP GTLEQCLNVD FCPQAARCCR TGVDEYGWIA AAVGWSLWFL TLILLCVDKL MKLTPDEPKD LQA (6-157 aa encoded by BC131697)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Transmembrane protein 213
Gene Aliases: TMEM213
UniProt ID: (Human) A2RRL7
Entrez Gene ID: (Human) 155006
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support