Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TMEM217 is a protein coding gene. Gene ontology (GO) annotation include fibrillar center; integral component of membrane; nucleolus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Transmembrane protein 217
Gene Aliases: C6orf128; dJ355M6.2; TMEM217
UniProt ID: (Human) Q8N7C4
Entrez Gene ID: (Human) 221468
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support