Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ASLIFIFTL FMVSCKVFLT ATHSHSSPFD ALEGYICPVL FGSACGGDHH HDNHGGSHSG GGPGAQHSAM PAKSKEELSE GSRKKKAKKA D (210-299 aa encoded by BC001195)
TMEM38A and TMEM38B are two recently identified trimeric intracellular cation (TRIC) channel subtypes. TMEM38A is preferentially expressed in excitable tissues such as striated muscle and brain and localizes to the sarcoplasmic reticulum (SR) in muscle tissues. Mice deficient in both TMEM38A and TMEM38B suffer embryonic cardiac failure; the cardiac myocytes display severe dysfunction in SR Ca2+ handling, weakened Ca2+ release, and reduced K+ permeability indicating that the TRIC cation channels are likely to act as counter-ion channels that function in synchronization with Ca2+ release from intracellular stores. Other experiments have shown that TMEM38A and TMEM38B can act with junctophilin proteins to support efficient ryanodine receptor-mediated Ca2+ release in muscle cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 27 kDa sarcoplasmic reticulum protein; Mitsugumin-33A; Mitsugumin33A; sarcoplasmic reticulum protein 27; SPR-27; tmem38a {ECO:0000250|UniProtKB:Q3TMP8}; Transmembrane protein 38A; TRIC-A; TRICA; Trimeric intracellular cation channel type A
Gene Aliases: 1110001E17Rik; AI413399; mg33a; RGD1307901; Srp-27; TMEM38A; TRIC-A; TRICA
UniProt ID: (Human) Q9H6F2, (Mouse) Q3TMP8, (Rat) A6ZIQ8
Entrez Gene ID: (Human) 79041, (Mouse) 74166, (Rat) 306327
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support