Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MAAPKGSLW VRTQLGLPPL LLLTMALAGG SGTASAEAFD SVLGDTASCH RACQLTYPLH TYPKEEELYA CQRGCRLFSI CQFVDDGIDL NRTKLECESA CTEAYSQSDE QYACHLGCQN QLPFAELRQE QLMSLMPKMH LLFPLTLVRS FWSDMMDSAQ SFITSSWTFY LQADDGKIVI FQSKPEIQYA PHLEQEPTNL RESSLSKMSS DLQMRNSQAH RNFLEDGESD GFLRCLSLNS GWILTTTLVL SVMVLLWICC ATVATAVEQY VPSEKLSIYG DLEFMNEQKL NRYPASSLVV VRSKTEDHEE AGPLPTKVNL AHSEI
Processing of the amyloid precursor protein (APP) by two different proteases, called alpha- and beta-secretase, is a central regulatory event in the generation of the amyloid beta peptide (Abeta), which has a key role in Alzheimer disease (AD) pathogenesis. TMEM59, a Golgi-localized protein, modulates the O-glycosylation and complex N-glycosylation steps occurring during the Golgi maturation of several proteins such as APP, BACE1, SEAP or PRNP. It inhibits APP transport and shedding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dendritic cell factor 1; Liver membrane-bound protein; RP4-758J24.2; Transmembrane protein 59
Gene Aliases: C1orf8; DCF1; HSPC001; PRO195; TMEM59; UNQ169; UNQ169/PRO195
UniProt ID: (Human) Q9BXS4
Entrez Gene ID: (Human) 9528
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support