Product References
LINC01123 promotes immune escape by sponging miR-214-3p to regulate B7-H3 in head and neck squamous-cell carcinoma.
Cell death & disease
Li H,Yang Z,Yang X,Zhang F,Wang J,Wu Z,Wanyan C,Meng Q,Gao W,Yang X,Wei J
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Thu Feb 03 00:00:00 EST 2022
PRDM1/BLIMP1 induces cancer immune evasion by modulating the USP22-SPI1-PD-L1 axis in hepatocellular carcinoma cells.
Nature communications
Li Q,Zhang L,You W,Xu J,Dai J,Hua D,Zhang R,Yao F,Zhou S,Huang W,Dai Y,Zhang Y,Baheti T,Qian X,Pu L,Xu J,Xia Y,Zhang C,Tang J,Wang X
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Mon Dec 12 00:00:00 EST 2022
Baricitinib treatment resolves lower-airway macrophage inflammation and neutrophil recruitment in SARS-CoV-2-infected rhesus macaques.
Cell
Hoang TN,Pino M,Boddapati AK,Viox EG,Starke CE,Upadhyay AA,Gumber S,Nekorchuk M,Busman-Sahay K,Strongin Z,Harper JL,Tharp GK,Pellegrini KL,Kirejczyk S,Zandi K,Tao S,Horton TR,Beagle EN,Mahar EA,Lee MYH,Cohen J,Jean SM,Wood JS,Connor-Stroud F,Stammen RL,Delmas OM,Wang S,Cooney KA,Sayegh MN,Wang L,Filev PD,Weiskopf D,Silvestri G,Waggoner J,Piantadosi A,Kasturi SP,Al-Shakhshir H,Ribeiro SP,Sekaly RP,Levit RD,Estes JD,Vanderford TH,Schinazi RF,Bosinger SE,Paiardini M
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Thu Jan 21 00:00:00 EST 2021
An AAV-based, room-temperature-stable, single-dose COVID-19 vaccine provides durable immunogenicity and protection in non-human primates.
Cell host & microbe
Zabaleta N,Dai W,Bhatt U,Hérate C,Maisonnasse P,Chichester JA,Sanmiguel J,Estelien R,Michalson KT,Diop C,Maciorowski D,Dereuddre-Bosquet N,Cavarelli M,Gallouët AS,Naninck T,Kahlaoui N,Lemaitre J,Qi W,Hudspeth E,Cucalon A,Dyer CD,Pampena MB,Knox JJ,LaRocque RC,Charles RC,Li D,Kim M,Sheridan A,Storm N,Johnson RI,Feldman J,Hauser BM,Contreras V,Marlin R,Tsong Fang RH,Chapon C,van der Werf S,Zinn E,Ryan A,Kobayashi DT,Chauhan R,McGlynn M,Ryan ET,Schmidt AG,Price B,Honko A,Griffiths A,Yaghmour S,Hodge R,Betts MR,Freeman MW,Wilson JM,Le Grand R,Vandenberghe LH
25-7349-41 was used in Flow Cytometry to show that AAVCOVID-1, an adeno-associated viral (AAV), spike-gene-based vaccine candidate demonstrates potent immunogenicity in mouse and non-human primates following a single injection and confers complete protection from SARS-CoV-2 challenge in macaques.
Wed Sep 08 00:00:00 EDT 2021
Membrane lymphotoxin-α2β is a novel tumor necrosis factor (TNF) receptor 2 (TNFR2) agonist.
Cell death & disease
Kucka K,Lang I,Zhang T,Siegmund D,Medler J,Wajant H
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Tue Apr 06 00:00:00 EDT 2021
NKD2 mediates stimulation-dependent ORAI1 trafficking to augment Ca2+ entry in T cells.
Cell reports
Wu B,Woo JS,Vila P,Jew M,Leung J,Sun Z,Srikanth S,Gwack Y
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Tue Aug 24 00:00:00 EDT 2021
Development of an Inflammatory CD14+ Dendritic Cell Subset in Humanized Mice.
Frontiers in immunology
Iwabuchi R,Ide K,Terahara K,Wagatsuma R,Iwaki R,Matsunaga H,Tsunetsugu-Yokota Y,Takeyama H,Takahashi Y
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Mon Sep 27 00:00:00 EDT 2021
Immunogenicity of an AAV-based, room-temperature stable, single dose COVID-19 vaccine in mice and non-human primates.
bioRxiv : the preprint server for biology
Zabaleta N,Dai W,Bhatt U,Chichester JA,Sanmiguel J,Estelien R,Michalson KT,Diop C,Maciorowski D,Qi W,Hudspeth E,Cucalon A,Dyer CD,Pampena MB,Knox JJ,LaRocque RC,Charles RC,Li D,Kim M,Sheridan A,Storm N,Johnson RI,Feldman J,Hauser BM,Zinn E,Ryan A,Kobayashi DT,Chauhan R,McGlynn M,Ryan ET,Schmidt AG,Price B,Honko A,Griffiths A,Yaghmour S,Hodge R,Betts MR,Freeman MW,Wilson JM,Vandenberghe LH
25-7349-41 was used in Flow Cytometry to show that this methodology holds as a very promising single dose, thermostable vaccine platform well-suited to address emerging pathogens on a global scale.
Tue Jan 19 00:00:00 EST 2021
Rejuvenating Effector/Exhausted CAR T Cells to Stem Cell Memory-Like CAR T Cells By Resting Them in the Presence of CXCL12 and the NOTCH Ligand.
Cancer research communications
Ando M,Kondo T,Tomisato W,Ito M,Shichino S,Srirat T,Mise-Omata S,Nakagawara K,Yoshimura A
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Fri Oct 01 00:00:00 EDT 2021
Inhibition of Cholesterol Esterification Enzyme Enhances the Potency of Human Chimeric Antigen Receptor T Cells against Pancreatic Carcinoma.
Molecular therapy oncolytics
Zhao L,Liu Y,Zhao F,Jin Y,Feng J,Geng R,Sun J,Kang L,Yu L,Wei Y
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Fri Mar 27 00:00:00 EDT 2020
Abrogation of HLA surface expression using CRISPR/Cas9 genome editing: a step toward universal T cell therapy.
Scientific reports
Lee J,Sheen JH,Lim O,Lee Y,Ryu J,Shin D,Kim YY,Kim M
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Tue Oct 20 00:00:00 EDT 2020
Mucosal-associated invariant T (MAIT) cells mediate protective host responses in sepsis.
eLife
Trivedi S,Labuz D,Anderson CP,Araujo CV,Blair A,Middleton EA,Jensen O,Tran A,Mulvey MA,Campbell RA,Hale JS,Rondina MT,Leung DT
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Mon Nov 09 00:00:00 EST 2020
CAIX Regulates GBM Motility and TAM Adhesion and Polarization through EGFR/STAT3 under Hypoxic Conditions.
International journal of molecular sciences
Huang BR,Liu YS,Lai SW,Lin HJ,Shen CK,Yang LY,Lu DY
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Fri Aug 14 00:00:00 EDT 2020
CAR T cells targeting tumor endothelial marker CLEC14A inhibit tumor growth.
JCI insight
Zhuang X,Maione F,Robinson J,Bentley M,Kaul B,Whitworth K,Jumbu N,Jinks E,Bystrom J,Gabriele P,Garibaldi E,Delmastro E,Nagy Z,Gilham D,Giraudo E,Bicknell R,Lee SP
25-7349-41 was used in Flow Cytometry to generate chimeric antigen receptors from 2 CLEC14A-specific antibodies and expressed them in T cells. In vitro studies demonstrated that, when exposed to their target antigen, these engineered T cells proliferate, release IFN-γ, and mediate cytotoxicity.
Fri Oct 02 00:00:00 EDT 2020
Vaccine induction of antibodies and tissue-resident CD8+ T cells enhances protection against mucosal SHIV-infection in young macaques.
JCI insight
Petitdemange C,Kasturi SP,Kozlowski PA,Nabi R,Quarnstrom CF,Reddy PBJ,Derdeyn CA,Spicer LM,Patel P,Legere T,Kovalenkov YO,Labranche CC,Villinger F,Tomai M,Vasilakos J,Haynes B,Kang CY,Gibbs JS,Yewdell JW,Barouch D,Wrammert J,Montefiori D,Hunter E,Amara RR,Masopust D,Pulendran B
25-7349-41 was used in Flow Cytometry to demonstrate that vaccination strategies that induce both CD8+ T cell and antibody responses can confer enhanced protection against infection.
Thu Feb 21 00:00:00 EST 2019
Heterogeneous Tumor-Immune Microenvironments among Differentially Growing Metastases in an Ovarian Cancer Patient.
Cell
Jiménez-Sánchez A,Memon D,Pourpe S,Veeraraghavan H,Li Y,Vargas HA,Gill MB,Park KJ,Zivanovic O,Konner J,Ricca J,Zamarin D,Walther T,Aghajanian C,Wolchok JD,Sala E,Merghoub T,Snyder A,Miller ML
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Thu Aug 24 00:00:00 EDT 2017
The distribution and function of human memory T cell subsets in lung cancer.
Immunologic research
Sheng SY,Gu Y,Lu CG,Zou JY,Hong H,Wang R
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Application of a whole blood mycobacterial growth inhibition assay to study immunity against Mycobacterium tuberculosis in a high tuberculosis burden population.
PloS one
Baguma R,Penn-Nicholson A,Smit E,Erasmus M,Day J,Makhethe L,de Kock M,Hughes EJ,van Rooyen M,Pienaar B,Stone L,Hanekom W,Brennan MJ,Wallis RS,Hatherill M,Scriba TJ
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Wed Oct 18 00:00:00 EDT 2017
IL-21 and probiotic therapy improve Th17 frequencies, microbial translocation, and microbiome in ARV-treated, SIV-infected macaques.
Mucosal immunology
Ortiz AM,Klase ZA,DiNapoli SR,Vujkovic-Cvijin I,Carmack K,Perkins MR,Calantone N,Vinton CL,Riddick NE,Gallagher J,Klatt NR,McCune JM,Estes JD,Paiardini M,Brenchley JM
25-7349 was used in Flow cytometry/Cell sorting to investigate methods to promote Th17 cell recovery in individuals infected with HIV, showing that IL-21 and probiotic therapy improve Th17 frequencies, microbial translocation, and microbiome in antiretroviral-treated, SIV-infected macaques.
Tue Mar 01 00:00:00 EST 2016
Long-lasting multifunctional CD8+ T cell responses in end-stage melanoma patients can be induced by dendritic cell vaccination.
Oncoimmunology
Wimmers F,Aarntzen EH,Duiveman-deBoer T,Figdor CG,Jacobs JF,Tel J,de Vries IJ
25-7349 was used in Flow cytometry/Cell sorting to investigate the functionality of tumour-specific CD8(+) T cells in melanoma patients upon dendritic cell vaccination, showing long-lasting multifunctional responses in end-stage patients.
Sat Jan 29 00:00:00 EST 2022
Macrophages Are Phenotypically and Functionally Diverse across Tissues in Simian Immunodeficiency Virus-Infected and Uninfected Asian Macaques.
Journal of virology
Ortiz AM,DiNapoli SR,Brenchley JM
25-7349 was used in Flow cytometry/Cell sorting to demonstrate that macrophages from healthy animals exhibit phenotypic and functional heterogeneity across tissues and across a variety of stimuli.
Mon Jun 01 00:00:00 EDT 2015
Qualification of a whole blood intracellular cytokine staining assay to measure mycobacteria-specific CD4 and CD8 T cell immunity by flow cytometry.
Journal of immunological methods
Kagina BM,Mansoor N,Kpamegan EP,Penn-Nicholson A,Nemes E,Smit E,Gelderbloem S,Soares AP,Abel B,Keyser A,Sidibana M,Hughes JE,Kaplan G,Hussey GD,Hanekom WA,Scriba TJ
25-7349 was used in Flow cytometry/Cell sorting to qualify the intracellular cytokine staining process and to assess the effects of long-term cryopreservation.
Sun Feb 01 00:00:00 EST 2015
Targeting aPKC disables oncogenic signaling by both the EGFR and the proinflammatory cytokine TNFα in glioblastoma.
Science signaling
Kusne Y,Carrera-Silva EA,Perry AS,Rushing EJ,Mandell EK,Dietrich JD,Errasti AE,Gibbs D,Berens ME,Loftus JC,Hulme C,Yang W,Lu Z,Aldape K,Sanai N,Rothlin CV,Ghosh S
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Tue Aug 12 00:00:00 EDT 2014
Monocyte subsets in schistosomiasis patients with periportal fibrosis.
Mediators of inflammation
Fernandes JS,Araujo MI,Lopes DM,de Souza Rda P,Carvalho EM,Cardoso LS
25-7349 was used in Flow cytometry/Cell sorting to suggest that the three different monocyte populations participate in the immunopathogenesis of periportal fibrosis.
Mon Dec 15 00:00:00 EST 2014
Cell-mediated immune responses to respiratory syncytial virus infection: magnitude, kinetics, and correlates with morbidity and age.
Human vaccines & immunotherapeutics
Geevarghese B,Weinberg A
Published figure using TNF alpha monoclonal antibody (Product # 25-7349-41) in Flow Cytometry
Mon Apr 25 00:00:00 EDT 2016
Functional OCT4-specific CD4+ and CD8+ T cells in healthy controls and ovarian cancer patients.
Oncoimmunology
Di J,Massuger LF,Duiveman-de Boer T,Zusterzeel PL,Figdor CG,Torensma R
25-7349 was used in Flow cytometry/Cell sorting to demonstrate the existence of anti-CSC specific T cells in ovarian cancer patients.
Wed May 01 00:00:00 EDT 2013
Cancer/testis antigens are novel targets of immunotherapy for adult T-cell leukemia/lymphoma.
Blood
Nishikawa H,Maeda Y,Ishida T,Gnjatic S,Sato E,Mori F,Sugiyama D,Ito A,Fukumori Y,Utsunomiya A,Inagaki H,Old LJ,Ueda R,Sakaguchi S
25-7349 was used in Flow cytometry/Cell sorting to investigate the possible expression of cancer/testis antigens by adult T-cell leukemia/lymphoma cells.
Thu Mar 29 00:00:00 EDT 2012
Brefeldin A, but not monensin, enables flow cytometric detection of interleukin-4 within peripheral T cells responding to ex vivo stimulation with Chlamydia trachomatis.
Journal of immunological methods
Vicetti Miguel RD,Maryak SA,Cherpes TL
25-7349 was used in Flow cytometry/Cell sorting to compare the ability of brefeldin A and monensin to concurrently trap 4 cytokines within peripheral blood mononuclear cells.
Wed Oct 31 00:00:00 EDT 2012
Differential expression of CD300a/c on human TH1 and TH17 cells.
BMC immunology
Simhadri VR,Mariano JL,Zhou Q,DeBell KE,Borrego F
25-7349 was used in Flow cytometry/Cell sorting to conclude that the CD300a/c receptors are differentially expressed on human TH1 and TH17 cells, and that their ligation is capable of modulating TCR mediated signals.
Wed Nov 02 00:00:00 EDT 2011
Human Th1 cells that express CD300a are polyfunctional and after stimulation up-regulate the T-box transcription factor eomesodermin.
PloS one
Narayanan S,Silva R,Peruzzi G,Alvarez Y,Simhadri VR,Debell K,Coligan JE,Borrego F
25-7349 was used in Flow cytometry/Cell sorting to investigate a subset within the CD4 T cell population defined by expression of the immunomodulatory receptor CD300a.
Thu May 13 00:00:00 EDT 2010
GM-CSF DNA: an adjuvant for higher avidity IgG, rectal IgA, and increased protection against the acute phase of a SHIV-89.6P challenge by a DNA/MVA immunodeficiency virus vaccine.
Virology
Lai L,Vödrös D,Kozlowski PA,Montefiori DC,Wilson RL,Akerstrom VL,Chennareddi L,Yu T,Kannanganat S,Ofielu L,Villinger F,Wyatt LS,Moss B,Amara RR,Robinson HL
25734941 was used in flow cytometry to assess the effects of using GM-CSF DNA with the DNA prime for a simian-human immunodeficiency virus-89.6 vaccine
Wed Dec 05 00:00:00 EST 2007