Product References
LINC01123 promotes immune escape by sponging miR-214-3p to regulate B7-H3 in head and neck squamous-cell carcinoma.
Cell death & disease
Li H,Yang Z,Yang X,Zhang F,Wang J,Wu Z,Wanyan C,Meng Q,Gao W,Yang X,Wei J
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Thu Feb 03 00:00:00 EST 2022
PRDM1/BLIMP1 induces cancer immune evasion by modulating the USP22-SPI1-PD-L1 axis in hepatocellular carcinoma cells.
Nature communications
Li Q,Zhang L,You W,Xu J,Dai J,Hua D,Zhang R,Yao F,Zhou S,Huang W,Dai Y,Zhang Y,Baheti T,Qian X,Pu L,Xu J,Xia Y,Zhang C,Tang J,Wang X
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Mon Dec 12 00:00:00 EST 2022
Baricitinib treatment resolves lower-airway macrophage inflammation and neutrophil recruitment in SARS-CoV-2-infected rhesus macaques.
Cell
Hoang TN,Pino M,Boddapati AK,Viox EG,Starke CE,Upadhyay AA,Gumber S,Nekorchuk M,Busman-Sahay K,Strongin Z,Harper JL,Tharp GK,Pellegrini KL,Kirejczyk S,Zandi K,Tao S,Horton TR,Beagle EN,Mahar EA,Lee MYH,Cohen J,Jean SM,Wood JS,Connor-Stroud F,Stammen RL,Delmas OM,Wang S,Cooney KA,Sayegh MN,Wang L,Filev PD,Weiskopf D,Silvestri G,Waggoner J,Piantadosi A,Kasturi SP,Al-Shakhshir H,Ribeiro SP,Sekaly RP,Levit RD,Estes JD,Vanderford TH,Schinazi RF,Bosinger SE,Paiardini M
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Thu Jan 21 00:00:00 EST 2021
Membrane lymphotoxin-α2β is a novel tumor necrosis factor (TNF) receptor 2 (TNFR2) agonist.
Cell death & disease
Kucka K,Lang I,Zhang T,Siegmund D,Medler J,Wajant H
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Tue Apr 06 00:00:00 EDT 2021
NKD2 mediates stimulation-dependent ORAI1 trafficking to augment Ca2+ entry in T cells.
Cell reports
Wu B,Woo JS,Vila P,Jew M,Leung J,Sun Z,Srikanth S,Gwack Y
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Tue Aug 24 00:00:00 EDT 2021
Development of an Inflammatory CD14+ Dendritic Cell Subset in Humanized Mice.
Frontiers in immunology
Iwabuchi R,Ide K,Terahara K,Wagatsuma R,Iwaki R,Matsunaga H,Tsunetsugu-Yokota Y,Takeyama H,Takahashi Y
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Mon Sep 27 00:00:00 EDT 2021
Rejuvenating Effector/Exhausted CAR T Cells to Stem Cell Memory-Like CAR T Cells By Resting Them in the Presence of CXCL12 and the NOTCH Ligand.
Cancer research communications
Ando M,Kondo T,Tomisato W,Ito M,Shichino S,Srirat T,Mise-Omata S,Nakagawara K,Yoshimura A
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Fri Oct 01 00:00:00 EDT 2021
Inhibition of Cholesterol Esterification Enzyme Enhances the Potency of Human Chimeric Antigen Receptor T Cells against Pancreatic Carcinoma.
Molecular therapy oncolytics
Zhao L,Liu Y,Zhao F,Jin Y,Feng J,Geng R,Sun J,Kang L,Yu L,Wei Y
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Fri Mar 27 00:00:00 EDT 2020
Abrogation of HLA surface expression using CRISPR/Cas9 genome editing: a step toward universal T cell therapy.
Scientific reports
Lee J,Sheen JH,Lim O,Lee Y,Ryu J,Shin D,Kim YY,Kim M
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Tue Oct 20 00:00:00 EDT 2020
Mucosal-associated invariant T (MAIT) cells mediate protective host responses in sepsis.
eLife
Trivedi S,Labuz D,Anderson CP,Araujo CV,Blair A,Middleton EA,Jensen O,Tran A,Mulvey MA,Campbell RA,Hale JS,Rondina MT,Leung DT
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Mon Nov 09 00:00:00 EST 2020
CAIX Regulates GBM Motility and TAM Adhesion and Polarization through EGFR/STAT3 under Hypoxic Conditions.
International journal of molecular sciences
Huang BR,Liu YS,Lai SW,Lin HJ,Shen CK,Yang LY,Lu DY
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Fri Aug 14 00:00:00 EDT 2020
Human Immunodeficiency Virus Infection Impairs Th1 and Th17 Mycobacterium tuberculosis-Specific T-Cell Responses.
The Journal of infectious diseases
Murray LW,Satti I,Meyerowitz J,Jones M,Willberg CB,Ussher JE,Goedhals D,Hurst J,Phillips RE,McShane H,Vuuren CV,Frater J
45-7349 was used in Flow cytometry/Cell sorting to measure expression of interferon gamma, tumor necrosis factor alpha, interleukin 2, and interleukin 17 in CD4+ and CD8+ T cells in response to Mtb antigens (PPD, ESAT-6/CFP-10 [EC], and DosR regulon-encoded α-crystallin [Rv2031c]).
Sat May 05 00:00:00 EDT 2018
Membrane-Associated Proteinase 3 on Granulocytes and Acute Myeloid Leukemia Inhibits T Cell Proliferation.
Journal of immunology (Baltimore, Md. : 1950)
Yang TH,St John LS,Garber HR,Kerros C,Ruisaard KE,Clise-Dwyer K,Alatrash G,Ma Q,Molldrem JJ
45-7349 was used in Flow cytometry/Cell sorting to investigate the effect of membrane P3-expressing polymorphonuclear neutrophils and acute myeloid leukaemia blasts on the proliferation of CD4 and CD8 T cells in vitro.
Sat Sep 01 00:00:00 EDT 2018
Schistosoma mansoni rSm29 Antigen Induces a Regulatory Phenotype on Dendritic Cells and Lymphocytes From Patients With Cutaneous Leishmaniasis.
Frontiers in immunology
Lopes DM,Oliveira SC,Page B,Carvalho LP,Carvalho EM,Cardoso LS
45-7349 was used in Flow cytometry/Cell sorting to evaluate the potential of the S.mansoni Sm29 antigen to change aspects of profiles of monocyte derived dendritic cells and lymphocytes from subjects with cutaneous leishmaniasis.
Tue Oct 15 00:00:00 EDT 2019
Heterogeneous Tumor-Immune Microenvironments among Differentially Growing Metastases in an Ovarian Cancer Patient.
Cell
Jiménez-Sánchez A,Memon D,Pourpe S,Veeraraghavan H,Li Y,Vargas HA,Gill MB,Park KJ,Zivanovic O,Konner J,Ricca J,Zamarin D,Walther T,Aghajanian C,Wolchok JD,Sala E,Merghoub T,Snyder A,Miller ML
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Thu Aug 24 00:00:00 EDT 2017
The distribution and function of human memory T cell subsets in lung cancer.
Immunologic research
Sheng SY,Gu Y,Lu CG,Zou JY,Hong H,Wang R
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Application of a whole blood mycobacterial growth inhibition assay to study immunity against Mycobacterium tuberculosis in a high tuberculosis burden population.
PloS one
Baguma R,Penn-Nicholson A,Smit E,Erasmus M,Day J,Makhethe L,de Kock M,Hughes EJ,van Rooyen M,Pienaar B,Stone L,Hanekom W,Brennan MJ,Wallis RS,Hatherill M,Scriba TJ
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Wed Oct 18 00:00:00 EDT 2017
Tissue factor expression by myeloid cells contributes to protective immune response against Mycobacterium tuberculosis infection.
European journal of immunology
Venkatasubramanian S,Tripathi D,Tucker T,Paidipally P,Cheekatla S,Welch E,Raghunath A,Jeffers A,Tvinnereim AR,Schechter ME,Andrade BB,Mackman N,Idell S,Vankayalapati R
45-7349 was used in Flow cytometry/Cell sorting to determine the role of tissue factor expressed by myeloid cells during Mycobacterium tuberculosis infection.
Mon Feb 01 00:00:00 EST 2016
Long-lasting multifunctional CD8+ T cell responses in end-stage melanoma patients can be induced by dendritic cell vaccination.
Oncoimmunology
Wimmers F,Aarntzen EH,Duiveman-deBoer T,Figdor CG,Jacobs JF,Tel J,de Vries IJ
45-7349 was used in Flow cytometry/Cell sorting to investigate the functionality of tumour-specific CD8(+) T cells in melanoma patients upon dendritic cell vaccination, showing long-lasting multifunctional responses in end-stage patients.
Sat Jan 29 00:00:00 EST 2022
PD-1+Tim-3+ CD8+ T Lymphocytes Display Varied Degrees of Functional Exhaustion in Patients with Regionally Metastatic Differentiated Thyroid Cancer.
Cancer immunology research
Severson JJ,Serracino HS,Mateescu V,Raeburn CD,McIntyre RC,Sams SB,Haugen BR,French JD
45-7349 was used in Flow cytometry/Cell sorting to determine that the release of PD-1 and Tim-3, and the inhibition of Tregs, may encourage tumour elimination in metastatic differentiated thyroid cancer.
Mon Jun 01 00:00:00 EDT 2015
The APPEESFRS Peptide, Restricted by the HLA-B*35:01 Molecule, and the APPEESFRF Variant Derived from an Autologous HIV-1 Strain Induces Polyfunctional Responses in CD8+ T Cells.
BioResearch open access
Acevedo-Sáenz L,Carmona-Pérez L,Velilla-Hernández PA,Delgado JC,Rugeles L MT
45-7349 was used in Flow cytometry/Cell sorting to determine the effector profile and maturation phenotype of CD8+ T-cells targeting the consensus APPEESFRS (AS9) epitope and its variant APPEESFRF (AF9).
Fri Aug 28 00:00:00 EDT 2015
Immunologic human renal allograft injury associates with an altered IL-10/TNF-α expression ratio in regulatory B cells.
Journal of the American Society of Nephrology : JASN
Cherukuri A,Rothstein DM,Clark B,Carter CR,Davison A,Hernandez-Fuentes M,Hewitt E,Salama AD,Baker RJ
45-7349 was used in Flow cytometry/Cell sorting to explore proinflammatory cytokine expression in B-cell subsets.
Tue Jul 01 00:00:00 EDT 2014
Targeting aPKC disables oncogenic signaling by both the EGFR and the proinflammatory cytokine TNFα in glioblastoma.
Science signaling
Kusne Y,Carrera-Silva EA,Perry AS,Rushing EJ,Mandell EK,Dietrich JD,Errasti AE,Gibbs D,Berens ME,Loftus JC,Hulme C,Yang W,Lu Z,Aldape K,Sanai N,Rothlin CV,Ghosh S
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Tue Aug 12 00:00:00 EDT 2014
Transcriptional and posttranscriptional regulation of cytokine gene expression in HIV-1 antigen-specific CD8+ T cells that mediate virus inhibition.
Journal of virology
Payne TL,Blackinton J,Frisbee A,Pickeral J,Sawant S,Vandergrift NA,Freel SA,Ferrari G,Keene JD,Tomaras GD
45-7349 was used in Flow cytometry/Cell sorting to show that mRNA stability, in addition to transcription, is key in regulating the direct anti-HIV-1 function of antigen-specific memory CD8+ T cells.
Mon Sep 01 00:00:00 EDT 2014
Cell-mediated immune responses to respiratory syncytial virus infection: magnitude, kinetics, and correlates with morbidity and age.
Human vaccines & immunotherapeutics
Geevarghese B,Weinberg A
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Mon Apr 25 00:00:00 EDT 2016
Functional OCT4-specific CD4+ and CD8+ T cells in healthy controls and ovarian cancer patients.
Oncoimmunology
Di J,Massuger LF,Duiveman-de Boer T,Zusterzeel PL,Figdor CG,Torensma R
Published figure using TNF alpha monoclonal antibody (Product # 45-7349-42) in Flow Cytometry
Wed May 01 00:00:00 EDT 2013
An unrestrained proinflammatory M1 macrophage population induced by iron impairs wound healing in humans and mice.
The Journal of clinical investigation
Sindrilaru A,Peters T,Wieschalka S,Baican C,Baican A,Peter H,Hainzl A,Schatz S,Qi Y,Schlecht A,Weiss JM,Wlaschek M,Sunderkötter C,Scharffetter-Kochanek K
45-7349 was used in Flow cytometry/Cell sorting to study human subjects with chronic venous leg ulcers and devise a human and mouse disease model for macrophage-driven chronic inflammation.
Tue Mar 01 00:00:00 EST 2011