Save up to 49% with protein expression workflow promo

Save now

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • TNF alpha Antibodies

            Invitrogen

            TNF alpha Monoclonal Antibody (MP6-XT22), PE, eBioscience™

            71 Published Figures
            101 References
            View all (221) TNF alpha antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 12-7321-41
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            143.00
            25 Tests
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite TNF alpha Monoclonal Antibody (MP6-XT22), PE, eBioscience™

            • Published Figures (71)
            Application
            TNF alpha Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.

            FIGURE: 1 / 71

            TNF alpha Antibody (12-7321-41) in Flow

            Nature immunology 2024 - Extended Data Fig. 2 Regulation of HB-EGF by astrocytes during autoimmune neuroinflammation. a , EAE development in wild-type mice with the timepoints analyzed (naive, peak, recovery, LSW) indicated. n = 20. b , tSNE plots of CNS cells (upper, downsampled to 30000 cells) and cell type abundance (% of singlets; c ) analyzed by high-dimensional flow cytometry. OLCs, oligodendrocyte lineage cells; OPCs, oligod... View More
            View Product

            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)
            TNF alpha Antibody in Flow Cytometry (Flow)

            Product Details

            12-7321-41

            Applications
            Tested Dilution
            Publications

            Immunohistochemistry (IHC)

            -
            View 1 publication 1 publication

            Flow Cytometry (Flow)

            5 µL (0.125 µg)/test
            View 99 publications 99 publications

            Functional Assay (Functional)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Mouse

            Published species

            Mammal, Mouse

            Host/Isotype

            Rat / IgG1, kappa

            Recommended Isotype Control

            Rat IgG1 kappa Isotype Control (eBRG1), PE, eBioscience™

            Class

            Monoclonal

            Type

            Antibody

            Clone

            MP6-XT22

            Conjugate

            PE
            • Unconjugated
            • Alexa Fluor 488 
            • Alexa Fluor 561
            • Alexa Fluor 660 
            • APC 
            • Brilliant Violet 421
            • Brilliant Violet 480
            • Brilliant Violet 605
            • Brilliant Violet 650
            • Brilliant Violet 786
            • eFluor 450
            • FITC
            • Functional Grade
            • PE-Cyanine7
            • PE-eFluor 610
            • PerCP-eFluor 710
            • Request custom conjugation

            Excitation/Emission Max

            565/576 nm View spectra spectra

            Form

            Liquid

            Concentration

            5 µL/Test

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2, with BSA

            Contains

            0.09% sodium azide

            Storage conditions

            4°C, store in dark, DO NOT FREEZE!

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_10854722

            Product Specific Information

            Description: The MP6-XT22 antibody reacts with mouse tumor necrosis factor-alpha (TNF alpha), a 17 kDa cytokine produced by monocytes, macrophages, neutrophils, NK cells and CD4(+) T cells. TNF alpha has cytolytic activity against a range of tumor cells and is important in immune regulation. TNF alpha forms dimers and trimers and also exists as a 26 kDa membrane-bound form.

            Applications Reported: This MP6-XT22 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

            Applications Tested: This MP6-XT22 antibody is tested by intracellular flow cytometric analysis of stimulated splenocytes. This product has been pre-titrated and tested by intracellular flow cytometric analysis. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

            Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            How to build a flow cytometry experiment with Invitrogen Flow Cytometry Panel Builder
            The Invitrogen Flow Cytometry Panel Builder webtool helps you chose antibody clones, fluorophores, fluorescent proteins, and reagents to design your experiment.
            Video Player is loading.
            Current Time 0:00
            /
            Duration 0:00
            Loaded: 0%
            0:00
            Stream Type LIVE
            Remaining Time -0:00
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected
            • en (Main), selected

            This is a modal window.

            Beginning of dialog window. Escape will cancel and close the window.

            Text
            Text Background
            Caption Area Background
            Font Size
            Text Edge Style
            Font Family

            End of dialog window.

            This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

            How to use the Panel Builder

            Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

            Bioinformatics

            Protein Aliases: Cachectin; DADB-70P7.1; TNF alpha; TNF superfamily; TNF α; TNF-a; TNF-alpha; TNFα; Tumor necrosis factor; Tumor necrosis factor ligand superfamily member 2; tumor necrosis factor-alpha

            View more View less

            Gene Aliases: DIF; Tnf; TNF-a; TNF-alpha; Tnfa; TNFalpha; Tnfsf1a; TNFSF2

            View more View less

            UniProt ID: (Mouse) P06804

            View more View less

            Entrez Gene ID: (Mouse) 21926

            View more View less

            Function(s)

            protease binding cytokine activity tumor necrosis factor receptor binding protein binding identical protein binding transcription regulatory region DNA binding

            Process(es)

            protein import into nucleus, translocation negative regulation of transcription from RNA polymerase II promoter MAPK cascade activation of MAPKKK activity activation of MAPK activity cell activation positive regulation of cytokine production regulation of protein phosphorylation positive regulation of protein phosphorylation negative regulation of L-glutamate transport chronic inflammatory response to antigenic stimulus negative regulation of cytokine secretion involved in immune response positive regulation of chronic inflammatory response to antigenic stimulus positive regulation of humoral immune response mediated by circulating immunoglobulin glucose metabolic process apoptotic process activation of cysteine-type endopeptidase activity involved in apoptotic process obsolete transformed cell apoptotic process defense response inflammatory response immune response humoral immune response signal transduction JNK cascade multicellular organism development cell proliferation positive regulation of cell proliferation negative regulation of cell proliferation extrinsic apoptotic signaling pathway via death domain receptors intrinsic apoptotic signaling pathway in response to DNA damage response to virus organ morphogenesis positive regulation of gene expression negative regulation of gene expression negative regulation of alkaline phosphatase activity calcium-mediated signaling extracellular matrix organization osteoclast differentiation sequestering of triglyceride positive regulation of protein complex assembly positive regulation of fever generation lipopolysaccharide-mediated signaling pathway negative regulation of interleukin-6 production positive regulation of chemokine production positive regulation of interferon-gamma production positive regulation of interleukin-18 production positive regulation of interleukin-6 production receptor biosynthetic process positive regulation of peptidyl-serine phosphorylation tumor necrosis factor-mediated signaling pathway positive regulation of heterotypic cell-cell adhesion regulation of cell proliferation positive regulation of NF-kappaB import into nucleus defense response to bacterium positive regulation of apoptotic process positive regulation of programmed cell death regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of protein complex disassembly positive regulation of protein complex disassembly positive regulation of cysteine-type endopeptidase activity involved in apoptotic process positive regulation of MAP kinase activity protein kinase B signaling positive regulation of JUN kinase activity positive regulation of neuron apoptotic process negative regulation of growth of symbiont in host negative regulation of viral genome replication positive regulation of chemokine biosynthetic process cellular extravasation positive regulation of interleukin-8 biosynthetic process positive regulation of nitric oxide biosynthetic process negative regulation of myoblast differentiation negative regulation of osteoblast differentiation regulation of osteoclast differentiation positive regulation of osteoclast differentiation positive regulation of action potential positive regulation of cell adhesion positive regulation of mitotic nuclear division positive regulation of protein kinase activity negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of translational initiation by iron negative regulation of glucose import positive regulation of JNK cascade positive regulation of smooth muscle cell proliferation regulation of protein secretion positive regulation of cytokine secretion positive regulation of inflammatory response positive regulation of phagocytosis regulation of insulin secretion positive regulation of synaptic transmission defense response to Gram-positive bacterium leukocyte migration leukocyte tethering or rolling detection of mechanical stimulus involved in sensory perception of pain negative regulation of lipid catabolic process regulation of immunoglobulin secretion positive regulation of membrane protein ectodomain proteolysis positive regulation of sequence-specific DNA binding transcription factor activity positive regulation of NF-kappaB transcription factor activity positive regulation of protein transport response to glucocorticoid positive regulation of NFAT protein import into nucleus positive regulation of hair follicle development positive regulation of protein kinase B signaling positive regulation of vitamin D biosynthetic process positive regulation of calcidiol 1-monooxygenase activity epithelial cell proliferation involved in salivary gland morphogenesis regulation of branching involved in salivary gland morphogenesis negative regulation of branching involved in lung morphogenesis cellular response to lipopolysaccharide cellular response to amino acid stimulus cellular response to nicotine cellular response to organic cyclic compound positive regulation of podosome assembly apoptotic signaling pathway extrinsic apoptotic signaling pathway necroptotic signaling pathway positive regulation of protein localization to cell surface positive regulation of chemokine (C-X-C motif) ligand 2 production regulation of reactive oxygen species metabolic process negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-4g5vm:80/100.66.76.159:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0