Product References
Immune signature of Chlamydia vaccine CTH522/CAF®01 translates from mouse-to-human and induces durable protection in mice.
Nature communications
Olsen AW,Rosenkrands I,Jacobsen CS,Cheeseman HM,Kristiansen MP,Dietrich J,Shattock RJ,Follmann F
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Feb 23 00:00:00 UTC 2024
The astrocyte-produced growth factor HB-EGF limits autoimmune CNS pathology.
Nature immunology
Linnerbauer M,Lößlein L,Vandrey O,Peter A,Han Y,Tsaktanis T,Wogram E,Needhamsen M,Kular L,Nagel L,Zissler J,Andert M,Meszaros L,Hanspach J,Zuber F,Naumann UJ,Diebold M,Wheeler MA,Beyer T,Nirschl L,Cirac A,Laun FB,Günther C,Winkler J,Bäuerle T,Jagodic M,Hemmer B,Prinz M,Quintana FJ,Rothhammer V
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2024
Neutrophil responsiveness to IL-10 impairs clearance of Streptococcus pneumoniae from the lungs.
Journal of leukocyte biology
Horn KJ,Fulte S,Yang M,Lorenz BP,Clark SE
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Jan 05 00:00:00 UTC 2024
An aging-related immune landscape in the hematopoietic immune system.
Immunity & ageing : I & A
Lv J,Zhang C,Liu X,Gu C,Liu Y,Gao Y,Huang Z,Jiang Q,Chen B,He D,Wang T,Xu Z,Su W
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Tue Jan 02 00:00:00 UTC 2024
Ly6C-high monocytes alleviate brain injury in experimental subarachnoid hemorrhage in mice.
Journal of neuroinflammation
Chen H,Xu C,Zeng H,Zhang Z,Wang N,Guo Y,Zheng Y,Xia S,Zhou H,Yu X,Fu X,Tang T,Wu X,Chen Z,Peng Y,Cai J,Li J,Yan F,Gu C,Chen G,Chen J
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Nov 17 00:00:00 UTC 2023
Tumor-secreted IFI35 promotes proliferation and cytotoxic activity of CD8+ T cells through PI3K/AKT/mTOR signaling pathway in colorectal cancer.
Journal of biomedical science
Li P,Zhou D,Chen D,Cheng Y,Chen Y,Lin Z,Zhang X,Huang Z,Cai J,Huang W,Lin Y,Ke H,Long J,Zou Y,Ye S,Lan P
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Wed Jun 28 00:00:00 UTC 2023
Conditional Knockout of Hypoxia-Inducible Factor 1-Alpha in Tumor-Infiltrating Neutrophils Protects against Pancreatic Ductal Adenocarcinoma.
International journal of molecular sciences
Sieow JL,Penny HL,Gun SY,Tan LQ,Duan K,Yeong JPS,Pang A,Lim D,Toh HC,Lim TKH,Engleman E,Rotzschke O,Ng LG,Chen J,Tan SM,Wong SC
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Sun Jan 01 00:00:00 UTC 2023
Infiltrating myeloid cell-derived properdin markedly promotes microglia-mediated neuroinflammation after ischemic stroke.
Journal of neuroinflammation
Liu PY,Li HQ,Dong MQ,Gu XY,Xu SY,Xia SN,Bao XY,Xu Y,Cao X
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Sat Nov 11 00:00:00 UTC 2023
Fc-γR-dependent antibody effector functions are required for vaccine-mediated protection against antigen-shifted variants of SARS-CoV-2.
Nature microbiology
Mackin SR,Desai P,Whitener BM,Karl CE,Liu M,Baric RS,Edwards DK,Chicz TM,McNamara RP,Alter G,Diamond MS
25-7321-82 was used in Flow cytometry/Cell sorting to suggest that Fc-FcγR engagement and alveolar macrophages are required for vaccine-induced antibody-mediated protection against infection by antigenically changed SARS-CoV-2 variants, including Omicron strains.
Sat Apr 01 00:00:00 UTC 2023
A single-amino acid substitution in the adaptor LAT accelerates TCR proofreading kinetics and alters T-cell selection, maintenance and function.
Nature immunology
Lo WL,Kuhlmann M,Rizzuto G,Ekiz HA,Kolawole EM,Revelo MP,Andargachew R,Li Z,Tsai YL,Marson A,Evavold BD,Zehn D,Weiss A
25-7321-82 was used in Immunocytochemistry to reveal the importance of kinetic proofreading in balancing tolerance and immunity.
Sat Apr 01 00:00:00 UTC 2023
Forsythiaside A Improves the Inhibitory Efficiency of Recombinant Protein Vaccines against Bovine Viral Diarrhea Virus Infection.
International journal of molecular sciences
Yang G,Wang J,Wang S,Zhu Y
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Sat Aug 20 00:00:00 UTC 2022
Cancer cell autophagy, reprogrammed macrophages, and remodeled vasculature in glioblastoma triggers tumor immunity.
Cancer cell
Chryplewicz A,Scotton J,Tichet M,Zomer A,Shchors K,Joyce JA,Homicsko K,Hanahan D
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Mon Oct 10 00:00:00 UTC 2022
Airway Prevotella promote TLR2-dependent neutrophil activation and rapid clearance of Streptococcus pneumoniae from the lung.
Nature communications
Horn KJ,Schopper MA,Drigot ZG,Clark SE
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Thu Jun 09 00:00:00 UTC 2022
Breast cancer cell-derived extracellular vesicles promote CD8+ T cell exhaustion via TGF-β type II receptor signaling.
Nature communications
Xie F,Zhou X,Su P,Li H,Tu Y,Du J,Pan C,Wei X,Zheng M,Jin K,Miao L,Wang C,Meng X,van Dam H,Ten Dijke P,Zhang L,Zhou F
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2022
Translation factor eIF5a is essential for IFNγ production and cell cycle regulation in primary CD8+ T lymphocytes.
Nature communications
Tan TCJ,Kelly V,Zou X,Wright D,Ly T,Zamoyska R
25-7321-82 was used in Flow cytometry/Cell sorting to show that the function of the elongation factor, eIF5a, is regulated dynamically in naive CD8+ T cells upon activation by post-translational modification, whereupon it facilitates translation of specific subsets of proteins.
Sat Dec 17 00:00:00 UTC 2022
Translation factor eIF5a is essential for IFNγ production and cell cycle regulation in primary CD8+ T lymphocytes.
Nature communications
Tan TCJ,Kelly V,Zou X,Wright D,Ly T,Zamoyska R
25-7321-82 was used in Flow cytometry/Cell sorting to show that the function of the elongation factor, eIF5a, is regulated dynamically in naive CD8+ T cells upon activation by post-translational modification, whereupon it facilitates translation of specific subsets of proteins.
Sat Dec 17 00:00:00 UTC 2022
Intranasal delivery of a small-molecule ErbB inhibitor promotes recovery from acute and late-stage CNS inflammation.
JCI insight
Linnerbauer M,Lößlein L,Vandrey O,Tsaktanis T,Beer A,Naumann UJ,Panier F,Beyer T,Nirschl L,Kuramatsu JB,Winkler J,Quintana FJ,Rothhammer V
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Apr 08 00:00:00 UTC 2022
Interleukin-37 protects against acinar cell pyroptosis in acute pancreatitis.
JCI insight
Ma N,Yuan C,Shi J,Zhu Q,Liu Y,Ma X,Li B,Gong W,Xue J,Lu G,Li W,Li J
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Tue Nov 08 00:00:00 UTC 2022
Circular RNA cancer vaccines drive immunity in hard-to-treat malignancies.
Theranostics
Li H,Peng K,Yang K,Ma W,Qi S,Yu X,He J,Lin X,Yu G
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Thu Sep 29 00:00:00 UTC 2022
Z. morio Hemolymph Relieves E. coli-Induced Mastitis by Inhibiting Inflammatory Response and Repairing the Blood-Milk Barrier.
International journal of molecular sciences
Zou Y,Wang X,Xu J,Wang S,Li S,Zhu Y,Wang J
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Mon Oct 31 00:00:00 UTC 2022
Z. morio Hemolymph Relieves E. coli-Induced Mastitis by Inhibiting Inflammatory Response and Repairing the Blood-Milk Barrier.
International journal of molecular sciences
Zou Y,Wang X,Xu J,Wang S,Li S,Zhu Y,Wang J
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Mon Oct 31 00:00:00 UTC 2022
Neoantigen cancer vaccine augments anti-CTLA-4 efficacy.
NPJ vaccines
Salvatori E,Lione L,Compagnone M,Pinto E,Conforti A,Ciliberto G,Aurisicchio L,Palombo F
25-7321-82 was used in Flow Cytometry to support the use of NCV delivered by DNA-EP with αCTLA-4 and suggest a new combined therapy for clinical testing.
Wed Feb 02 00:00:00 UTC 2022
Human poliovirus receptor contributes to biliary atresia pathogenesis by exacerbating natural-killer-cell-mediated bile duct injury.
Liver international : official journal of the International Association for the Study of the Liver
Li Y,Li TY,Qi Q,Zhang MT,Tong MX,Su PJ,Zhang ZB
25-7321-82 was used in Flow cytometry/Cell sorting to show that poliovirus receptor contributes to BA pathogenesis by regulating NK cell-mediated bile duct injury; PVR has the value as a biomarker of BA.
Thu Dec 01 00:00:00 UTC 2022
COVID-19 treatments and pathogenesis including anosmia in K18-hACE2 mice.
Nature
Zheng J,Wong LR,Li K,Verma AK,Ortiz ME,Wohlford-Lenane C,Leidinger MR,Knudson CM,Meyerholz DK,McCray PB,Perlman S
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2021
The ubiquitin ligase MDM2 sustains STAT5 stability to control T cell-mediated antitumor immunity.
Nature immunology
Zhou J,Kryczek I,Li S,Li X,Aguilar A,Wei S,Grove S,Vatan L,Yu J,Yan Y,Liao P,Lin H,Li J,Li G,Du W,Wang W,Lang X,Wang W,Wang S,Zou W
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2021
Mapping and role of T cell response in SARS-CoV-2-infected mice.
The Journal of experimental medicine
Zhuang Z,Lai X,Sun J,Chen Z,Zhang Z,Dai J,Liu D,Li Y,Li F,Wang Y,Zhu A,Wang J,Yang W,Huang J,Li X,Hu L,Wen L,Zhuo J,Zhang Y,Chen D,Li S,Huang S,Shi Y,Zheng K,Zhong N,Zhao J,Zhou D,Zhao J
25-7321 was used in Flow cytometry/Cell sorting to demonstrate cross-reactive T cell responses between SARS-CoV and SARS-CoV-2, but not MERS-CoV, in mice.
Mon Apr 05 00:00:00 UTC 2021
Mapping and role of T cell response in SARS-CoV-2-infected mice.
The Journal of experimental medicine
Zhuang Z,Lai X,Sun J,Chen Z,Zhang Z,Dai J,Liu D,Li Y,Li F,Wang Y,Zhu A,Wang J,Yang W,Huang J,Li X,Hu L,Wen L,Zhuo J,Zhang Y,Chen D,Li S,Huang S,Shi Y,Zheng K,Zhong N,Zhao J,Zhou D,Zhao J
25-7321 was used in Flow cytometry/Cell sorting to demonstrate cross-reactive T cell responses between SARS-CoV and SARS-CoV-2, but not MERS-CoV, in mice.
Mon Apr 05 00:00:00 UTC 2021
Plant-derived exosomal microRNAs inhibit lung inflammation induced by exosomes SARS-CoV-2 Nsp12.
Molecular therapy : the journal of the American Society of Gene Therapy
Teng Y,Xu F,Zhang X,Mu J,Sayed M,Hu X,Lei C,Sriwastva M,Kumar A,Sundaram K,Zhang L,Park JW,Chen SY,Zhang S,Yan J,Merchant ML,Zhang X,McClain CJ,Wolfe JK,Adcock RS,Chung D,Palmer KE,Zhang HG
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Wed Aug 04 00:00:00 UTC 2021
TNF-Like Ligand 1 Aberrance Aggravates Nonalcoholic Steatohepatitis via M1 Macrophage Polarization.
Oxidative medicine and cellular longevity
Luo Y,Guo J,Jia W,Wu M,Yin F,Niu G,Shih DQ,Targan SR,Zhang X
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Mar 04 00:00:00 UTC 2022
Prevention of EAE by tolerogenic vaccination with PEGylated antigenic peptides.
Therapeutic advances in chronic disease
Pfeil J,Simonetti M,Lauer U,von Thülen B,Durek P,Poulsen C,Pawlowska J,Kröger M,Krähmer R,Leenders F,Hoffmann U,Hamann A
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Tue Nov 07 00:00:00 UTC 2023
M2 macrophage-derived extracellular vesicles facilitate CD8+T cell exhaustion in hepatocellular carcinoma via the miR-21-5p/YOD1/YAP/β-catenin pathway.
Cell death discovery
Pu J,Xu Z,Nian J,Fang Q,Yang M,Huang Y,Li W,Ge B,Wang J,Wei H
25-7321-82 was used in Flow Cytometry to demonstrate the involvement of M2 macrophage-derived extracellular vesicles in hepatocellular carcinoma-associated T cell exhaustion.
Fri Jul 16 00:00:00 UTC 2021
M2 macrophage-derived extracellular vesicles facilitate CD8+T cell exhaustion in hepatocellular carcinoma via the miR-21-5p/YOD1/YAP/β-catenin pathway.
Cell death discovery
Pu J,Xu Z,Nian J,Fang Q,Yang M,Huang Y,Li W,Ge B,Wang J,Wei H
25-7321-82 was used in Flow Cytometry to demonstrate the involvement of M2 macrophage-derived extracellular vesicles in hepatocellular carcinoma-associated T cell exhaustion.
Fri Jul 16 00:00:00 UTC 2021
T cell-derived tumor necrosis factor induces cytotoxicity by activating RIPK1-dependent target cell death.
JCI insight
Chun N,Ang RL,Chan M,Fairchild RL,Baldwin WM,Horwitz JK,Gelles JD,Chipuk JE,Kelliher MA,Pavlov VI,Li Y,Homann D,Heeger PS,Ting AT
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Wed Dec 22 00:00:00 UTC 2021
USP12 downregulation orchestrates a protumourigenic microenvironment and enhances lung tumour resistance to PD-1 blockade.
Nature communications
Yang Z,Xu G,Wang B,Liu Y,Zhang L,Jing T,Tang M,Xu X,Jiao K,Xiang L,Fu Y,Tang D,Zhang X,Jin W,Zhuang G,Zhao X,Liu Y
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Wed Aug 11 00:00:00 UTC 2021
Type 1 Cytotoxic T Cells Increase in Placenta after Intrauterine Inflammation.
Frontiers in immunology
Liu J,Liu Y,Panda S,Liu A,Lei J,Burd I
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Dec 17 00:00:00 UTC 2021
Mechanisms that allow vaccination against an oncolytic vesicular stomatitis virus-encoded transgene to enhance safety without abrogating oncolysis.
Scientific reports
AuYeung AWK,Mould RC,Stegelmeier AA,van Vloten JP,Karimi K,Woods JP,Petrik JJ,Wood GA,Bridle BW
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Tue Jul 27 00:00:00 UTC 2021
The RNase MCPIP3 promotes skin inflammation by orchestrating myeloid cytokine response.
Nature communications
Liu B,Huang J,Ashraf A,Rahaman O,Lou J,Wang L,Cai P,Wen J,Anwaar S,Liu X,Ni H,Ganguly D,Zhao J,Yang CY
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Jul 02 00:00:00 UTC 2021
DA-DRD5 signaling controls colitis by regulating colonic M1/M2 macrophage polarization.
Cell death & disease
Liu L,Wu Y,Wang B,Jiang Y,Lin L,Li X,Yang S
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Mon May 17 00:00:00 UTC 2021
GPR120 induces regulatory dendritic cells by inhibiting HK2-dependent glycolysis to alleviate fulminant hepatic failure.
Cell death & disease
Yu H,Yang W,Huang J,Miao X,Wang B,Ren X,Gu Y,Wang Q,Ding X,Guo X,Qian F,Zhang Y,Xu H,Zheng L,Jin M
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Thu Dec 16 00:00:00 UTC 2021
Characterisation of Macrophage Polarisation in Mice Infected with Ninoa Strain of Trypanosoma cruzi.
Pathogens (Basel, Switzerland)
Medina-Buelvas DM,Rodríguez-Sosa M,Vega L
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Sat Nov 06 00:00:00 UTC 2021
Antitumor efficacy of a neoantigen cancer vaccine delivered by electroporation is influenced by microbiota composition.
Oncoimmunology
Lione L,Salvatori E,Petrazzuolo A,Massacci A,Maggio R,Confroti A,Compagnone M,Aurisicchio L,Ciliberto G,Palombo F
25-7321-82 was used in Flow cytometry/Cell sorting to find that decreased microbiota diversity induced by prolonged antibiotic (ATB) treatment is associated with higher intratumor specific immune responses and consequently to a better antitumor effect induced by neoantigen cancer vaccines (NCVs) delivered by DNA-EP.
Tue Mar 23 00:00:00 UTC 2021
Rate of Freeze Impacts the Survival and Immune Responses Post Cryoablation of Melanoma.
Frontiers in immunology
Yakkala C,Dagher J,Sempoux C,Chiang CL,Denys A,Kandalaft LE,Koppolu B,Duran R
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Mon Dec 13 00:00:00 UTC 2021
PTENα functions as an immune suppressor and promotes immune resistance in PTEN-mutant cancer.
Nature communications
Sun Y,Lu D,Yin Y,Song J,Liu Y,Hao W,Qi F,Zhang G,Zhang X,Liu L,Lin Z,Liang H,Zhao X,Jin Y,Yin Y
25-7321-82 was used in Flow Cytometry to propose that PTENα protects tumor from T cell killing and thus PTENα is a potential target in antitumor immunotherapy.
Thu Aug 26 00:00:00 UTC 2021
Metabolic requirements of NK cells during the acute response against retroviral infection.
Nature communications
Littwitz-Salomon E,Moreira D,Frost JN,Choi C,Liou KT,Ahern DK,O'Shaughnessy S,Wagner B,Biron CA,Drakesmith H,Dittmer U,Finlay DK
25-7321-82 was used in Flow cytometry/Cell sorting to show that following infection natural killer cells increase nutrient uptake, including amino acids and iron, and reprogram their metabolic machinery by increasing glycolysis and mitochondrial metabolism.
Fri Sep 10 00:00:00 UTC 2021
A distal enhancer at risk locus 11q13.5 promotes suppression of colitis by Treg cells.
Nature
Nasrallah R,Imianowski CJ,Bossini-Castillo L,Grant FM,Dogan M,Placek L,Kozhaya L,Kuo P,Sadiyah F,Whiteside SK,Mumbach MR,Glinos D,Vardaka P,Whyte CE,Lozano T,Fujita T,Fujii H,Liston A,Andrews S,Cozzani A,Yang J,Mitra S,Lugli E,Chang HY,Unutmaz D,Trynka G,Roychoudhuri R
25-7321 was used in Flow cytometry/Cell sorting to provide a mechanistic basis for association of the 11q13.5 risk locus with immune-mediated diseases and identify GARP as a potential target in their therapy.
Wed Jul 01 00:00:00 UTC 2020
Developmental Relationships of Four Exhausted CD8+ T Cell Subsets Reveals Underlying Transcriptional and Epigenetic Landscape Control Mechanisms.
Immunity
Beltra JC,Manne S,Abdel-Hakeem MS,Kurachi M,Giles JR,Chen Z,Casella V,Ngiow SF,Khan O,Huang YJ,Yan P,Nzingha K,Xu W,Amaravadi RK,Xu X,Karakousis GC,Mitchell TC,Schuchter LM,Huang AC,Wherry EJ
25-7321-82 was used in Flow cytometry/Cell sorting to reveal a four-stage developmental hierarchy for Tex cells and define the molecular, transcriptional, and epigenetic mechanisms that could provide opportunities to improve cancer immunotherapy.
Tue May 19 00:00:00 UTC 2020
Fluvastatin-Pretreated Donor Cells Attenuated Murine aGVHD by Balancing Effector T Cell Distribution and Function under the Regulation of KLF2.
BioMed research international
Zhao K,Tian Y,Wang J,Chen C,Pan B,Yan Z,Zhu S,Xu K
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Sep 10 00:00:00 UTC 2021
Synthetic DNA Vaccines Adjuvanted with pIL-33 Drive Liver-Localized T Cells and Provide Protection from Plasmodium Challenge in a Mouse Model.
Vaccines
Reeder SM,Reuschel EL,Bah MA,Yun K,Tursi NJ,Kim KY,Chu J,Zaidi FI,Yilmaz I,Hart RJ,Perrin B,Xu Z,Humeau L,Weiner DB,Aly ASI
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Jan 10 00:00:00 UTC 2020
Deletion of immune evasion genes provides an effective vaccine design for tumor-associated herpesviruses.
NPJ vaccines
Brar G,Farhat NA,Sukhina A,Lam AK,Kim YH,Hsu T,Tong L,Lin WW,Ware CF,Blackman MA,Sun R,Wu TT
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Thu Nov 05 00:00:00 UTC 2020
Mucosal-associated invariant T cells promote inflammation and intestinal dysbiosis leading to metabolic dysfunction during obesity.
Nature communications
Toubal A,Kiaf B,Beaudoin L,Cagninacci L,Rhimi M,Fruchet B,da Silva J,Corbett AJ,Simoni Y,Lantz O,Rossjohn J,McCluskey J,Lesnik P,Maguin E,Lehuen A
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Jul 24 00:00:00 UTC 2020
Dissecting the phenotypic and functional heterogeneity of mouse inflammatory osteoclasts by the expression of Cx3cr1.
eLife
Madel MB,Ibáñez L,Ciucci T,Halper J,Rouleau M,Boutin A,Hue C,Duroux-Richard I,Apparailly F,Garchon HJ,Wakkach A,Blin-Wakkach C
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Wed May 13 00:00:00 UTC 2020
High-Throughput CRISPR Screening Identifies Genes Involved in Macrophage Viability and Inflammatory Pathways.
Cell reports
Covarrubias S,Vollmers AC,Capili A,Boettcher M,Shulkin A,Correa MR,Halasz H,Robinson EK,O'Briain L,Vollmers C,Blau J,Katzman S,McManus MT,Carpenter S
25-7321-82 was used in Flow Cytometry to unravel complexities of the TNF signaling cascade, showing that it can function in an autocrine manner in macrophages to negatively regulate the pathway.
Tue Dec 29 00:00:00 UTC 2020
Modulation of Vaccine-Induced HIV-1-Specific Immune Responses by Co-Electroporation of PD-L1 Encoding DNA.
Vaccines
Tannig P,Peter AS,Lapuente D,Klessing S,Damm D,Tenbusch M,Überla K,Temchura V
25-7321-82 was used in Flow Cytometry to indicate that co-electroporation of soluble checkpoint ectodomains together with DNA-based vaccines has modulatory effects on vaccine-induced immune responses that could improve vaccine efficacies.
Tue Jan 14 00:00:00 UTC 2020
Tolerogenic Immunomodulation by PEGylated Antigenic Peptides.
Frontiers in immunology
Pfeil J,Simonetti M,Lauer U,Volkmer R,von Thülen B,Durek P,Krähmer R,Leenders F,Hamann A,Hoffmann U
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Thu Apr 29 00:00:00 UTC 2021
Tpl2 Protects Against Fulminant Hepatitis Through Mobilization of Myeloid-Derived Suppressor Cells.
Frontiers in immunology
Xu J,Pei S,Wang Y,Liu J,Qian Y,Huang M,Zhang Y,Xiao Y
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Wed Oct 14 00:00:00 UTC 2020
Aging alters the immunological response to ischemic stroke.
Acta neuropathologica
Ritzel RM,Lai YJ,Crapser JD,Patel AR,Schrecengost A,Grenier JM,Mancini NS,Patrizz A,Jellison ER,Morales-Scheihing D,Venna VR,Kofler JK,Liu F,Verma R,McCullough LD
25-7321 was used in Flow Cytometry to study the marked differences in the composition of circulating and infiltrating leukocytes recruited to the ischaemic brain of old male mice after stroke compared to young male mice.
Sun Jul 01 00:00:00 UTC 2018
Interleukin 1 receptor (IL-1R1) activation exacerbates toxin-induced acute kidney injury.
American journal of physiology. Renal physiology
Privratsky JR,Zhang J,Lu X,Rudemiller N,Wei Q,Yu YR,Gunn MD,Crowley SD
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Sat Sep 01 00:00:00 UTC 2018
Targeting macrophage and microglia activation with colony stimulating factor 1 receptor inhibitor is an effective strategy to treat injury-triggered neuropathic pain.
Molecular pain
Lee S,Shi XQ,Fan A,West B,Zhang J
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Mon Oct 22 00:00:00 UTC 2018
Regulation of chitinase-3-like-1 in T cell elicits Th1 and cytotoxic responses to inhibit lung metastasis.
Nature communications
Kim DH,Park HJ,Lim S,Koo JH,Lee HG,Choi JO,Oh JH,Ha SJ,Kang MJ,Lee CM,Lee CG,Elias JA,Choi JM
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Mon Feb 05 00:00:00 UTC 2018
1,25-Dihydroxyvitamin D suppresses M1 macrophages and promotes M2 differentiation at bone injury sites.
JCI insight
Wasnik S,Rundle CH,Baylink DJ,Yazdi MS,Carreon EE,Xu Y,Qin X,Lau KW,Tang X
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Thu Sep 06 00:00:00 UTC 2018
Cell-Intrinsic Glycogen Metabolism Supports Early Glycolytic Reprogramming Required for Dendritic Cell Immune Responses.
Cell metabolism
Thwe PM,Pelgrom LR,Cooper R,Beauchamp S,Reisz JA,D'Alessandro A,Everts B,Amiel E
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Tue Sep 05 00:00:00 UTC 2017
A CCR4 antagonist reverses the tumor-promoting microenvironment of renal cancer.
The Journal of clinical investigation
Berlato C,Khan MN,Schioppa T,Thompson R,Maniati E,Montfort A,Jangani M,Canosa M,Kulbe H,Hagemann UB,Duncan AR,Fletcher L,Wilkinson RW,Powles T,Quezada SA,Balkwill FR
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2017
Egr2 and 3 control adaptive immune responses by temporally uncoupling expansion from T cell differentiation.
The Journal of experimental medicine
Miao T,Symonds ALJ,Singh R,Symonds JD,Ogbe A,Omodho B,Zhu B,Li S,Wang P
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Mon Jun 05 00:00:00 UTC 2017
A New Adjuvant MTOM Mediates Mycobacterium tuberculosis Subunit Vaccine to Enhance Th1-Type T Cell Immune Responses and IL-2+ T Cells.
Frontiers in immunology
Yu Q,Wang X,Fan X
Wed Nov 20 00:00:00 UTC 2019
Betulin from Hedyotis hedyotidea ameliorates concanavalin A-induced and T cell-mediated autoimmune hepatitis in mice.
Acta pharmacologica Sinica
Zhou YQ,Weng XF,Dou R,Tan XS,Zhang TT,Fang JB,Wu XW
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Wed Feb 01 00:00:00 UTC 2017
Tristetraprolin inhibits macrophage IL-27-induced activation of antitumour cytotoxic T cell responses.
Nature communications
Wang Q,Ning H,Peng H,Wei L,Hou R,Hoft DF,Liu J
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Wed Oct 11 00:00:00 UTC 2017
The TLR7 agonist imiquimod induces anti-cancer effects via autophagic cell death and enhances anti-tumoral and systemic immunity during radiotherapy for melanoma.
Oncotarget
Cho JH,Lee HJ,Ko HJ,Yoon BI,Choe J,Kim KC,Hahn TW,Han JA,Choi SS,Jung YM,Lee KH,Lee YS,Jung YJ
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Tue Apr 11 00:00:00 UTC 2017
Single-cell profiling reveals GPCR heterogeneity and functional patterning during neuroinflammation.
JCI insight
Tischner D,Grimm M,Kaur H,Staudenraus D,Carvalho J,Looso M,Günther S,Wanke F,Moos S,Siller N,Breuer J,Schwab N,Zipp F,Waisman A,Kurschus FC,Offermanns S,Wettschureck N
Thu Aug 03 00:00:00 UTC 2017
Antibiotic-induced perturbations in microbial diversity during post-natal development alters amyloid pathology in an aged APPSWE/PS1ΔE9 murine model of Alzheimer's disease.
Scientific reports
Minter MR,Hinterleitner R,Meisel M,Zhang C,Leone V,Zhang X,Oyler-Castrillo P,Zhang X,Musch MW,Shen X,Jabri B,Chang EB,Tanzi RE,Sisodia SS
25-7321 was used in Flow cytometry/Cell sorting to investigate how ABX-induced microbial diversity perturbations during post-natal stages of development coincide with altered host immunity mechanisms and amyloidosis in a murine model of AD.
Tue Sep 05 00:00:00 UTC 2017
Scramblase TMEM16F terminates T cell receptor signaling to restrict T cell exhaustion.
The Journal of experimental medicine
Hu Y,Kim JH,He K,Wan Q,Kim J,Flach M,Kirchhausen T,Vortkamp A,Winau F
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Mon Nov 14 00:00:00 UTC 2016
Reprogramming the immunological microenvironment through radiation and targeting Axl.
Nature communications
Aguilera TA,Rafat M,Castellini L,Shehade H,Kariolis MS,Hui AB,Stehr H,von Eyben R,Jiang D,Ellies LG,Koong AC,Diehn M,Rankin EB,Graves EE,Giaccia AJ
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Dec 23 00:00:00 UTC 2016
A Genome-wide CRISPR Screen in Primary Immune Cells to Dissect Regulatory Networks.
Cell
Parnas O,Jovanovic M,Eisenhaure TM,Herbst RH,Dixit A,Ye CJ,Przybylski D,Platt RJ,Tirosh I,Sanjana NE,Shalem O,Satija R,Raychowdhury R,Mertins P,Carr SA,Zhang F,Hacohen N,Regev A
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Thu Jul 30 00:00:00 UTC 2015
Single-Dose CpG Immunization Protects Against a Heterosubtypic Challenge and Generates Antigen-Specific Memory T Cells.
Frontiers in immunology
Vogel AJ,Brown DM
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Jul 10 00:00:00 UTC 2015
Enhancing the Quality of Antibodies to HIV-1 Envelope by GagPol-Specific Th Cells.
Journal of immunology (Baltimore, Md. : 1950)
Storcksdieck genannt Bonsmann M,Niezold T,Temchura V,Pissani F,Ehrhardt K,Brown EP,Osei-Owusu NY,Hannaman D,Hengel H,Ackerman ME,Streeck H,Nabi G,Tenbusch M,Überla K
25-7321 was used in Flow cytometry/Cell sorting to enhance the quality of antibodies to HIV-1 envelope by GagPol-Specific T helper Cells.
Sun Nov 15 00:00:00 UTC 2015
Immunogenicity and protective efficacy of DMT liposome-adjuvanted tuberculosis subunit CTT3H vaccine.
Human vaccines & immunotherapeutics
Teng X,Tian M,Li J,Tan S,Yuan X,Yu Q,Jing Y,Zhang Z,Yue T,Zhou L,Fan X
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Fri Mar 11 00:00:00 UTC 2016
Triclosan Induces Thymic Stromal Lymphopoietin in Skin Promoting Th2 Allergic Responses.
Toxicological sciences : an official journal of the Society of Toxicology
Marshall NB,Lukomska E,Long CM,Kashon ML,Sharpnack DD,Nayak AP,Anderson KL,Jean Meade B,Anderson SE
25-7321 was used in Flow cytometry/Cell sorting to examine the effects of dermal triclosan exposure on the skin and lymph nodes of mice and in a human skin model to identify mechanisms for augmenting allergic responses.
Tue Sep 01 00:00:00 UTC 2015
Nizatidine, a small molecular compound, enhances killed H5N1 vaccine cell-mediated responses and protects mice from lethal viral challenge.
Human vaccines & immunotherapeutics
Wang S,Wu B,Xue J,Wang M,Chen R,Wang B
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Mon Jan 05 00:00:00 UTC 2015
Helminth-induced interleukin-4 abrogates invariant natural killer T cell activation-associated clearance of bacterial infection.
Infection and immunity
Hsieh YJ,Fu CL,Hsieh MH
25-7321 was used in Flow cytometry/Cell sorting to reveal a previously unappreciated role for helminth-induced IL-4 in impairment of iNKT cell-mediated clearance of bacterial coexposure.
Thu May 01 00:00:00 UTC 2014
Evaluation of Lassa virus vaccine immunogenicity in a CBA/J-ML29 mouse model.
Vaccine
Goicochea MA,Zapata JC,Bryant J,Davis H,Salvato MS,Lukashevich IS
Published figure using TNF alpha monoclonal antibody (Product # 25-7321-82) in Flow Cytometry
Tue Feb 14 00:00:00 UTC 2012