Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ILIDRCGKH FGTILNYLRD DTITLPQNRQ EIKELMAEAK YYLIQGLVNM CQSALQDKKD SYQPVCNIPI ITSLKEEERL IESSTKPVVK LLYNRSNNKY SYTSNSDDHL LKNIELFDKL SLRFNGRVLF IKDVIGDEIC CWSFYGQGRK LAEVCCTSIV YATEKKQTKV (71-239 aa encoded by BC001949)
This gene was identified as a gene whose expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. Studies of a similar gene in mouse suggest that the expression of this gene is developmentally regulated in a tissue-specific manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 2; BTB/POZ domain-containing protein TNFAIP1; hBACURD2; Protein B12; tumor necrosis factor, alpha induced protein 1; tumor necrosis factor, alpha-induced protein 1 (endothelial); Tumor necrosis factor, alpha-induced protein 1, endothelial
Gene Aliases: B12; B61; BACURD2; BTBD34; EDP1; hBACURD2; TNFAIP1
UniProt ID: (Human) Q13829
Entrez Gene ID: (Human) 7126
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support