Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: ADQKRGKKQT AREMKKKILA ERRKPLNIDH LGEDKLRDKA KELWETLHQL EIDKFEFGEK LKRQKYDITT LRSRIDQAQK HSKKAGTPAK GKVGGRWK
Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta TnTF; Beta-TnTF; Fast skeletal muscle troponin T; fTnT; TnTf; troponin T type 3 (skeletal, fast); Troponin T, fast skeletal muscle; troponin-T3, skeletal, fast
Gene Aliases: TNNT3; TNTF
UniProt ID: (Human) P45378
Entrez Gene ID: (Human) 7140
Molecular Function:
actin binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support