Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TNPO1 comprises the beta subunit of the karyopherin receptor complex which interacts with nuclear localization signals to target nuclear proteins to the nucleus. The karyopherin receptor complex is a heterodimer of an alpha subunit which recognizes the nuclear localization signal and a beta subunit which docks the complex at nucleoporins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: importin 2; importin beta 2; Importin beta-2; karyopherin (importin) beta 2; Karyopherin beta-2; M9 region interaction protein; MAPKAP1; MIP; MSIN1; SAPK 1; SIN-1; SIN1; SIN1g; Transportin-1
Gene Aliases: AU021749; D13Ertd688e; IPO2; KPNB2; MIP; MIP1; TNPO1; TRN
UniProt ID: (Human) Q92973, (Mouse) Q8BFY9
Entrez Gene ID: (Rabbit) 100343169, (Human) 3842, (Mouse) 238799
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support