Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody recognizes a region near the N-terminus of TNS1.
Tensin is involved in the maintenance of cellular structure by anchoring Actin filaments at the focal adhesion via F-Actin binding and capping activities. However, tensin also contains a Src homology 2 (SH2) domain and has the ability to be phosphorylated. Tensin is phosphorylated on tyrosine, serine, and threonine residues, suggesting that it might participate in signal transduction cascades. These diverse characteristics in a single molecule indicate that tensin may be an important link between the cytoskeleton and signal transduction pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp586K0617; matrix-remodelling associated 6; Matrix-remodelling-associated protein 6; MGC88584; protein phosphatase 1, regulatory subunit 155; tensin 1; Tensin-1
Gene Aliases: 1110018I21Rik; 1200014E20Rik; AI648117; E030018G17Rik; E030037J05Rik; MST091; MST122; MST127; MSTP091; MSTP122; MSTP127; MXRA6; PPP1R155; TNS; TNS1
UniProt ID: (Human) Q9HBL0, (Mouse) E9Q0S6
Entrez Gene ID: (Human) 7145, (Mouse) 21961
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support