Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • TOMM40 Antibodies

            Proteintech

            TOMM40 Polyclonal Antibody

            79 References
            View all (15) TOMM40 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 18409-1-AP
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            492.00
            150 µL
             
            In stock
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite TOMM40 Polyclonal Antibody

            • Antibody Testing Data (17)
            Application
            TOMM40 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 17

            TOMM40 Antibody (18409-1-AP) in ICC/IF

            Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using TOMM40 antibody (Product # 18409-1-AP) at dilution of 1:400 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L), (CL594-Phalloidin, red).
            View Product

            TOMM40 Antibody in Immunocytochemistry (ICC/IF)
            TOMM40 Antibody in Immunocytochemistry (ICC/IF)
            TOMM40 Antibody in Immunocytochemistry (ICC/IF)
            TOMM40 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            TOMM40 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            TOMM40 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            TOMM40 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            TOMM40 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            TOMM40 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            TOMM40 Antibody in Western Blot (WB)
            TOMM40 Antibody in Western Blot (WB)
            TOMM40 Antibody in Western Blot (WB)
            TOMM40 Antibody in Western Blot (WB)
            TOMM40 Antibody in Western Blot (WB)
            TOMM40 Antibody in Western Blot (WB)
            TOMM40 Antibody in Flow Cytometry (Flow)
            TOMM40 Antibody in Immunoprecipitation (IP)

            Product Details

            18409-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:2400
            View 61 publications 61 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:100-1:400
            -

            Immunocytochemistry (ICC/IF)

            1:10-1:100
            View 13 publications 13 publications

            Flow Cytometry (Flow)

            0.2 µg/1x10^6 cells
            -

            Immunoprecipitation (IP)

            0.5-4.0 µg
            View 3 publications 3 publications

            Not applicable (N/A)

            -
            View 2 publications 2 publications
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            TOMM40 Fusion Protein Ag13065 (1-361 aa encoded by BC017224)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.22 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: MGNVLAASS PPAGPPPPPA PALVGLPPPP PSPPGFTLPP LGGSLGAGTS TSRSSERTPG AATASASGAA EDGACGCLPN PGTFEECHRK CKELFPIQME GVKLTVNKGL SNHFQVNHTV ALSTIGESNY HFGVTYVGTK QLSPTEAFPV LVGDMDNSGS LNAQVIHQLG PGLRSKMAIQ TQQSKFVNWQ VDGEYRGSDF TAAVTLGNPD VLVGSGILVA HYLQSITPCL ALGGELVYHR RPGEEGTVMS LAGKYTLNNW LATVTLGQAG MHATYYHKAS DQLQVGVEFE ASTRMQDTSV SFGYQLDLPK ANLLFKGSVD SNWIVGATLE KKLPPLPLTL ALGAFLNHRK NKFQCGFGLT IG (1-361 aa encoded by BC017224)

            Target Information

            TOMM40 is the channel-forming subunit of the translocase of the mitochondrial outer membrane complex that is essential for protein import into mitochondria.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: haymaker protein; Mitochondrial import receptor subunit TOM40 homolog; mitochondrial outer membrane protein; mitochondrial outer membrane protein 35; Mitochondrial outer membrane protein of 35 kDa; Mitochondrial outer membrane protein of 38 kDa; MOM35; OM38; outer membrane protein 38 kDa; p38.5; PER EC1; Protein Haymaker; Translocase of outer membrane 40 kDa subunit homolog; translocase of outer mitochondrial membrane 40 homolog

            View more View less

            Gene Aliases: AW539759; C19orf1; D19S1177E; Mom35; OM38; PER-EC1; PEREC1; TOM40; Tom40a; TOMM40

            View more View less

            UniProt ID: (Human) O96008, (Rat) Q75Q40, (Mouse) Q9QYA2

            View more View less

            Entrez Gene ID: (Human) 10452, (Rat) 308416, (Mouse) 53333

            View more View less

            Function(s)

            protein transmembrane transporter activity protein channel activity porin activity primary active transporter

            Process(es)

            protein targeting to mitochondrion ion transport macroautophagy protein import into mitochondrial matrix protein transport transmembrane transport protein transmembrane transport transport
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-rms4r:80/100.66.77.126:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0