Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
TOMM40 is the channel-forming subunit of the translocase of the mitochondrial outer membrane complex that is essential for protein import into mitochondria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: haymaker protein; Mitochondrial import receptor subunit TOM40 homolog; mitochondrial outer membrane protein; mitochondrial outer membrane protein 35; Mitochondrial outer membrane protein of 35 kDa; Mitochondrial outer membrane protein of 38 kDa; MOM35; OM38; outer membrane protein 38 kDa; p38.5; PER EC1; Protein Haymaker; Translocase of outer membrane 40 kDa subunit homolog; translocase of outer mitochondrial membrane 40 homolog
Gene Aliases: AW539759; C19orf1; D19S1177E; Mom35; OM38; PER-EC1; PEREC1; TOM40; Tom40a; TOMM40
UniProt ID: (Human) O96008, (Rat) Q75Q40, (Mouse) Q9QYA2
Entrez Gene ID: (Human) 10452, (Rat) 308416, (Mouse) 53333
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support