Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAASKPVEAA VVAAAVPSSG SGVGGGGTAG PGTGGLPRWQ LALAVGAPLL LGAGAIYLWS RQQRRREARG RGDASGLKRN SERKTPEGRA SPAPGSGHPE GPGAHLDMNS LDRAQAAKNK GNKYFKAGKY EQAIQCYTEA ISLCPTEKNV DLSTFYQNRA AAFEQLQKWK EVAQDCTKAV ELNPKYVKAL FRRAKAHEKL DNKKECLEDV TAVCILEGFQ NQQSMLLADK VLKLLGKEKA KEKYKNREPL MPSPQFIKSY FSSFTDDIIS QPMLKGEKSD EDKDKEGEAL EVKENSGYLK AKQYMEEENY DKIISECSKE IDAEGKYMAE ALLLRATFYL LIGNANAAKP DLDKVISLKE ANVKLRANAL IKRGSMYMQQ QQPLLSTQDF NMAADIDPQN ADVYHHRGQL KILLDQVEEA VADFDECIRL RPESALAQAQ KCFALYRQAY TGNNSSQIQA AMKGFEEVIK KFPRCAEGYA LYAQALTDQQ QFGKADEMYD KCIDLEPDNA TTYVHKGLLQ LQWKQDLDRG LELISKAIEI DNKCDFAYET MGTIEVQRGN MEKAIDMFNK AINLAKSEME MAHLYSLCDA AHAQTEVAKK YGLKPPTL
Functional mitochondria require up to 1000 proteins to function properly, with 99% synthesized as precursors in the cytoplasm and transported into the mitochondria with the aid of cytosolic chaperones and mitochondrial translocators (import components). Proteins to be imported are chaperoned to the mitochondria by the cytosolic heat shock protein (cHSP70) and are immediately pursued by Translocators of the Outer Membrane (TOMs), followed by transient interactions of the unfolded proteins with Translocators of the Inner Membrane (TIMs). TOMM70A is ubiquitously expressed in human tissues and localizes in the mitochondria. TOMM70A could play a significant role in the import of nuclear-encoded mitochondrial proteins with internal targeting sites such as ADP/ATP carriers and the uncoupling proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ90470; Mitochondrial import receptor subunit TOM70; Mitochondrial precursor proteins import receptor; Translocase of outer membrane 70 kDa subunit; translocase of outer mitochondrial membrane 70 homolog A; Translocase of outer mitochondrial membrane protein 70
Gene Aliases: KIAA0719; TOM70; TOMM70; TOMM70A
UniProt ID: (Human) O94826
Entrez Gene ID: (Human) 9868
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support