Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IAGVFPQKFD GDSAYVGMSD GNPELLSTSQ TYNGQSENNE DYEIPPITPP N
Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 80%.
TOX2 (TOX High Mobility Group Box Family Member 2) is a Protein Coding gene. Diseases associated with TOX2 include Conduct Disorder. Gene Ontology (GO) annotations related to this gene include chromatin binding and RNA polymerase II transcription factor binding. An important paralog of this gene is TOX.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GCX-1; granulosa cell HMG box 1; Granulosa cell HMG box protein 1; TOX high mobility group box family member 2
Gene Aliases: C20orf100; dJ1108D11.2; dJ495O3.1; GCX-1; GCX1; TOX2
UniProt ID: (Human) A8K1J1
Entrez Gene ID: (Human) 84969
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support