Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Peptide sequence: AGQKASAAFS SVGSVITKKL EDVKNSPTFK SFEEKVENLK SKVGGTKPAG
Sequence homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%
It belongs to the TPD52 family. Its exact function remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mD52; prostate and colon associated protein; prostate leucine zipper; Protein N8; Tumor protein D52
Gene Aliases: AI043038; D52; hD52; mD52; N8L; PC-1; PrLZ; TPD52
UniProt ID: (Human) P55327, (Mouse) Q62393
Entrez Gene ID: (Human) 7163, (Mouse) 21985
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support