Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 7
Recommended positive controls: 293T, A431, HeLa, HepG2, Neuro2A, GL261, C8D30, NIH-3T3, BCL-1, Raw264.7, C2C12, PC-12, Rat2.
Predicted reactivity: Mouse (100%), Rat (100%), Dog (100%), Chimpanzee (100%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate and dihydroxy-acetone phosphate in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 49; Methylglyoxal synthase; MGC88108; TIM; Triose-phosphate isomerase; Triosephosphate isomerase
Gene Aliases: AI255506; HEL-S-49; TIM; TPI; Tpi-1; TPI1; TPID
UniProt ID: (Human) P60174, (Rat) P48500, (Mouse) P17751
Entrez Gene ID: (Human) 7167, (Rat) 24849, (Mouse) 21991
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support