Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
IA-2 (insulinoma-associated protein 2, ICA512, PTPRN) is a member of the transmembrane protein tyrosine phosphatase (PTP) family located in secretory granules of neuroendocrine cells. The IA-2 protein is 979 amino acids in length and consists of a luminal domain, transmembrane domain, and cytoplasmic domain. Although a member of the PTP family, IA-2 lacks phosphatase activity with known substrates due to amino acid substitutions at critical sites in its PTP domain. Two paralog RPTPs, IA-2 and IA-2beta were identified as major autoantigens in type-1 diabetes mellitus. On granule exocytosis, the IA-2 cytoplasmic domain is cleaved and the resulting cytosolic fragment moves into the nucleus where it enhances the levels of phosphorylated STAT5 and STAT3, thereby inducing insulin gene transcription and granule biogenesis. IA-2 signaling enhances pancreatic beta-cell proliferation by regulating cyclins D through STATs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 105 kDa islet cell antigen; BEM-3; Brain-enriched membrane-associated protein tyrosine phosphatase; ICA 512; ICA105; ICA512; islet cell antigen 2; Islet cell antigen 512; Islet cell autoantigen 3; protein tyrosine phosphatase, receptor type, N; protein tyrosine phosphatase, receptor-type, N; protein tyrosine phosphatase-like N; PTP IA-2; PTPLP; R-PTP-N; Receptor-type tyrosine-protein phosphatase-like N
Gene Aliases: IA-2; IA-2/PTP; IA2; ICA3; ICA512; mIA-A; mKIAA4064; PTPRN; R-PTP-N
UniProt ID: (Human) Q16849, (Rat) Q63259
Entrez Gene ID: (Human) 5798, (Rat) 116660, (Mouse) 19275
Molecular Function:
protein phosphatase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support