Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: MAGSSSLEAV RRKIRSLQEQ ADAAEERAGT LQRELDHERK LRETAEADVA SLNRRIQLVE EELDRAQERL ATALQKLEEA EKAADESERG MKVIESRAQK DEEKMEIQEI QLKEAKHIAE DADRKYEEVA RKLVIIESDL ERAEERAELS EGKCAELEEE LKTVTNNLKS LEAQAEKYSQ KEDRYEEEIK VLSDKLKEAE TRAEFAERSV TKLEKSIDDL EDQLYQQLEQ NRRLTNELKL ALNED; Positive Samples: A-549, BT-474, THP-1, DU145, SW480, MCF7, Mouse heart, Mouse skeletal muscle, Mouse brain; Cellular Location: Cytoplasm, cytoskeleton
Tropomyosin is a dimeric coiled-coil protein that binds along the length of actin filaments. It is associated with the thin filaments of muscle cells and the microfilaments of nonmuscle cells. Chicken embryo fibroblasts (CEF) contain five isoforms of tropomyosin (a, b, 1, 2, and 3), identified as such by their different apparent molecular masses after separation by SDS-PAGE, but similar biochemical properties, such as resistance to heat and organic solvents, the ability to bind to F-actin filaments, and the lack of proline and tryptophan.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha tropomyosin; Alpha-tropomyosin; cardiomyopathy, hypertrophic 3; epididymis secretory protein Li 265; sarcomeric tropomyosin kappa; TPM1-alpha; tropomyosin; Tropomyosin alpha-1 chain; Tropomyosin-1
Gene Aliases: AA986836; AI854628; alpha-TM; C15orf13; CMD1Y; CMH3; HEL-S-265; HTM-alpha; LVNC9; TM2; Tm3; Tmpa; TMSA; Tpm-1; TPM1; Tpma
UniProt ID: (Human) P09493, (Mouse) P58771
Entrez Gene ID: (Human) 7168, (Mouse) 22003
Molecular Function: actin binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support