Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TPP-2; TPP-II; Tripeptidyl aminopeptidase; tripeptidyl peptidase II; Tripeptidyl-peptidase 2; Tripeptidyl-peptidase II
Gene Aliases: TPP-2; TPP2; TPPII
UniProt ID: (Human) P29144
Entrez Gene ID: (Human) 7174
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support