Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RRPPHPLPPR LTIHAPPQEL GPPVQRIQMT RRQSVGRGLQ LTPGIGGMQQ HFFDDEDRTV PSTPTLVVPH RTDGFAEAIH SPQVAGVPRF RFGPPEDMPQ TSSSHS
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Steroidogenic factor-1 (SF-1) regulates multiple genes involved in the adrenal and gonadal development and in the biosynthesis of a variety of hormones, including adrenal and gonadal steroids, anti-Mullerian hormone (AMH), and gonadotropins. SF-1 belongs to the fushi tarazu factor-1 (FTZ-F1) subfamily of orphan nuclear receptors. In the adult ovary, SF-1 localizes to theca/interstitial cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Megator; NPC-associated intranuclear protein; nuclear pore complex-associated protein TPR; Nucleoprotein TPR; translocated promoter region (to activated MET oncogene); Translocated promoter region protein; tumor potentiating region
Gene Aliases: TPR
UniProt ID: (Human) P12270
Entrez Gene ID: (Human) 7175
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support