Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQLTHQLDL FPECRVTLLL FKDVKNAGDL RRKAMEGTID GSLINPTVIV DPFQILVAAN KAVHLYKLGK MKTRTLSTEI IFNLSPNNNI SEALKKFGIS ANDTSILIVY IEEGEKQINQ EYLISQVEGH QVSLKNLPEI MNITEVKKIY KLSSQEESIG TLLDAIICRM STKDVL (1-175 aa encoded by B C029492)
TPRKB is a component of the EKC/KEOPS complex that is required for the formation of a threonylcarbamoyl group on adenosine at position 37 (t6A37) in tRNAs that read codons beginning with adenine. The complex is probably involved in the transfer of the threonylcarbamoyl moiety of threonylcarbamoyl-AMP (TC-AMP) to the N6 group of A37. TPRKB acts as an allosteric effector that regulates the t6A activity of the complex. TPRKB is not required for tRNA modification.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EKC/KEOPS complex subunit TPRKB; PRPK (p53-related protein kinase)-binding protein; PRPK-binding protein; TP53RK-binding protein; WBSCR18
Gene Aliases: CGI-121; CGI121; My019; TPRKB
UniProt ID: (Human) Q9Y3C4
Entrez Gene ID: (Human) 51002
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support