Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: SVLDNITTDK ILIDVFDGLP LYDDVKVQFF YSNLPTYYDN CSFYFWLHTS FIENNRLYLP KNELDNLHKQ KARRIYPSDF AVEILFGEKM TSSDVVAGSD
TPTE encodes a putative transmembrane tyrosine phosphatase that may be involved in signal transduction pathways of the endocrine or spermatogenetic function of the testis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 44; CT44; PTEN-related tyrosine phosphatase; Putative tyrosine-protein phosphatase TPTE; tensin, putative protein-tyrosine phosphatase; Transmembrane phosphatase with tensin homology; Tumor antigen BJ-HCC-5
Gene Aliases: CT44; PTEN2; TPTE
UniProt ID: (Human) P56180
Entrez Gene ID: (Human) 7179
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support