Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MASSSGSSPR PAPDENEFPF GCPPTVCQDP KEPRALCCAG CLSENPRNGE DQICPKCRGE DLQSISPGSR LRTQEKAHPE VAEAGIGCPF AGVGCSFKGS PQSVQEHEVT SQTSHLNLLL GFMKQWKARL GCGLESGPMA LEQNLSDLQL QAAVEVAGDL EVDCYRAPCS ESQEELALQH FMKEKLLAEL EGKLRVFENI VAVLNKEVEA SHLALATSIH QSQLDRERIL SLEQRVVELQ QTLAQKDQAL GKLEQSLRLM EEASFDGTFL WKITNVTRRC HESACGRTVS LFSPAFYTAK YGYKLCLRLY LNGDGTGKRT HLSLFIVIMR GEYDALLPWP FRNKVTFMLL DQNNREHAID AFRPDLSSAS FQRPQSETNV ASGCPLFFPL SKLQSPKHAY VKDDTMFLKC IVETST
The protein encoded by this gene is a member of the TNF receptor (TNFR) associated factor (TRAF) protein family. TRAF proteins associate with, and mediate the signal transduction from various receptors of the TNFR superfamily. This protein and TRAF2 form a heterodimeric complex, which is required for TNF-alpha-mediated activation of MAPK8/JNK and NF-kappaB. The protein complex formed by this protein and TRAF2 also interacts with inhibitor-of-apoptosis proteins (IAPs), and thus mediates the anti-apoptotic signals from TNF receptors. The expression of this protein can be induced by Epstein-Barr virus (EBV). EBV infection membrane protein 1 (LMP1) is found to interact with this and other TRAF proteins; this interaction is thought to link LMP1-mediated B lymphocyte transformation to the signal transduction from TNFR family receptors. Three transcript variants encoding two different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epstein-Bar virus-induced protein 6; Epstein-Barr virus-induced protein 6; TNF receptor-associated factor 1
Gene Aliases: EBI6; MGC:10353; TRAF1
UniProt ID: (Human) Q13077
Entrez Gene ID: (Human) 7185
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support