Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSQSQNAIF TSPTGEENLM NSNHRDSESI TDVCSNEDLP EVELVSLLEE QLPQYRLKVD TLFLYENQDW TQSPHQRQHA SDALSPVLAE ETFRYMILGT DRVEQMTKTY NDIDMVTHLL AERDRDLELA ARIGQALLKR NHILSEQNES LEEQLGQAFD QVNQLQHELC KKDELLRIVS IASEESETDS SCSTPLRFNE SFSLSQGLLQ LEMLQEKLKE LEEENMALRS KACHIKTETV TYEEKEQQLV SDCVKELRET NAQMSRMTEE LSGKSDELVR YQEELSSLLS QIVDLQHKLK EHVIEKEELK LHLQASKDAQ RQLTMELHEL QDRNMECLGM LHESQEEIKE LRSRS (1-354 aa encoded by BC040668)
May regulate endosome-to-lysosome trafficking of membrane cargo, including EGFR.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3 homolog; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein homolog; EMBL AAH60681.1; GABA-A receptor interacting factor-1; GABA-A receptor-interacting factor 1; gamma-aminobutyric acid(A) receptor-interacting factor; GRIF-1; milton homolog 2; O-GlcNAc transferase-interacting protein of 98 kDa; O-linked N-acetylglucosamine transferase interacting protein 98; Trafficking kinesin-binding protein 2; trafficking protein, kinesin binding 2
Gene Aliases: 2900022D04Rik; 4733401O11Rik; AI480836; ALS2CR3; AW046271; CALS-C; GRIF-1; GRIF1; KIAA0549; MILT2; mKIAA0549; OIP98; TRAK2
UniProt ID: (Human) O60296, (Rat) Q8R2H7
Entrez Gene ID: (Human) 66008, (Mouse) 70827, (Rat) 171086
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support