Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MREGIVTAT EQEVKEDIAK LLRYESSALP SGQLTSLSEY ASRMRAGTRN IYYLCAPNRH LAEHSPYYEA MKKKDTEVLF CFEQFDELTL LHLREFDKKK LISVETDIVV DHYKEEKFED RSPAAECLSE KETEELMAWM RNVLGSRVTN VKVTLRLDTH PAMVTVLEMG AARHFLRMQQ LAKTQEERAQ LLQPTLEINP RHALIKKLNQ LRASEPGLAQ LLVDQIYENA MIAAGLVDDP RAMVGRLNEL LVKALERH (446-704 aa encoded by BC000406)
The 90 kDa heat shock protein (hsp90) family of molecular chaperones is a highly conserved family of proteins that play an important physiological role. Hsp90 is involved in numerous cellular processes but is best known for its association with signal transduction machinery. A recently cloned homolog of hsp90 is the tumor necrosis factor receptor-associated protein (TRAP1). Like hsp90, TRAP1 is found to be associated with numerous proteins involved in diverse actions. Immunofluorescence data has shown TRAP1 to be localized in the mitochondria of mammalian cells. This observation and the fact that TRAP1 is shown to have a mitochondrial targeting presequence strongly implicates TRAP1 as a mitochondrial matrix protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heat shock protein 75 kDa, mitochondrial; HSP 75; testicular tissue protein Li 209; TNF receptor-associated protein 1; TNFR-associated protein 1; TRAP-1; tumor necrosis factor type 1 receptor associated protein; Tumor necrosis factor type 1 receptor-associated protein
Gene Aliases: 2410002K23Rik; HSP 75; HSP75; HSP90L; TRAP-1; TRAP1
UniProt ID: (Human) Q12931, (Rat) Q5XHZ0, (Mouse) Q9CQN1
Entrez Gene ID: (Human) 10131, (Rat) 287069, (Mouse) 68015
Molecular Function:
Hsp90 family chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support